missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N,N',N'-Tetramethylethylenediamine 98.0+%, TCI America™

$32.65 - $127.14

Chemical Identifiers

CAS 110-18-9
Molecular Formula C6H16N2
Molecular Weight (g/mol) 116.208
MDL Number MFCD00008335
InChI Key KWYHDKDOAIKMQN-UHFFFAOYSA-N
Synonym temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl
PubChem CID 8037
ChEBI CHEBI:32850
IUPAC Name N,N,N',N'-tetramethylethane-1,2-diamine
SMILES CN(C)CCN(C)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T014725ML
View Documents
TCI America
T014725ML
25 mL N/A
 
T0147100ML
View Documents
TCI America
T0147100ML
100 mL N/A
 
T0147500ML
View Documents
TCI America
T0147500ML
500 mL N/A
 
Description

Description

Specifications

Chemical Identifiers

110-18-9
116.208
KWYHDKDOAIKMQN-UHFFFAOYSA-N
8037
N,N,N',N'-tetramethylethane-1,2-diamine
C6H16N2
MFCD00008335
temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl
CHEBI:32850
CN(C)CCN(C)C

Specifications

110-18-9
Colorless
C6H16N2
25 mL
temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl
CN(C)CCN(C)C
116.208
CHEBI:32850
≥98.0% (GC,T)
N,N,N′,N′-Tetramethylethylenediamine
-55°C
122°C
MFCD00008335
2372
KWYHDKDOAIKMQN-UHFFFAOYSA-N
N,N,N',N'-tetramethylethane-1,2-diamine
8037
116.21
Liquid
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0155

RTECSNumber : KV7175000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'