missing translation for 'onlineSavingsMsg'
Learn More

N,N,N'-Trimethylethylenediamine, 97%

CAS: 142-25-6 | C5H14N2 | 102.18 g/mol

$133.23 - $133.23

Chemical Identifiers

CAS 142-25-6
Molecular Formula C5H14N2
Molecular Weight (g/mol) 102.18
MDL Number MFCD00014874
InChI Key HVOYZOQVDYHUPF-UHFFFAOYSA-N
Synonym n,n,n'-trimethylethylenediamine, n1,n1,n2-trimethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n'-trimethyl, 2-dimethylamino ethyl methyl amine, n,n,n-trimethylethylenediamine, ethylenediamine, n,n,n'-trimethyl, n1,n1,n2-trimethyl-1,2-ethanediamine, 1,2-ethanediamine, n1,n1,n2-trimethyl, dimethyl 2-methylamino ethyl amine, n,n,n'-trimethylethane-1,2-diamine
PubChem CID 67338
IUPAC Name N,N',N'-trimethylethane-1,2-diamine
SMILES CNCCN(C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC366910050
SDS View Documents
Thermo Scientific Chemicals
366910050
5 g Glass bottle
Each for $133.23
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

142-25-6
102.18
HVOYZOQVDYHUPF-UHFFFAOYSA-N
67338
CNCCN(C)C
C5H14N2
MFCD00014874
n,n,n'-trimethylethylenediamine, n1,n1,n2-trimethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n'-trimethyl, 2-dimethylamino ethyl methyl amine, n,n,n-trimethylethylenediamine, ethylenediamine, n,n,n'-trimethyl, n1,n1,n2-trimethyl-1,2-ethanediamine, 1,2-ethanediamine, n1,n1,n2-trimethyl, dimethyl 2-methylamino ethyl amine, n,n,n'-trimethylethane-1,2-diamine
N,N',N'-trimethylethane-1,2-diamine

Specifications

142-25-6
100.0
0.8000g/mL
9°C
96% min. (GC)
C5H14N2
(CH3)2NCH2CH2NHCH3
5 g
0.8
HVOYZOQVDYHUPF-UHFFFAOYSA-N
N,N',N'-trimethylethane-1,2-diamine
67338
97%
N, N, N'-Trimethylethylenediamine
96.0
Colorless
116°C to 118°C
Authentic
Glass bottle
1.4185 to 1.4205
MFCD00014874
04, I, 415
n,n,n'-trimethylethylenediamine, n1,n1,n2-trimethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n'-trimethyl, 2-dimethylamino ethyl methyl amine, n,n,n-trimethylethylenediamine, ethylenediamine, n,n,n'-trimethyl, n1,n1,n2-trimethyl-1,2-ethanediamine, 1,2-ethanediamine, n1,n1,n2-trimethyl, dimethyl 2-methylamino ethyl amine, n,n,n'-trimethylethane-1,2-diamine
CNCCN(C)C
102.18
102.18
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful in contact with skin.
Causes serious eye damage.
Highly flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

EINECSNumber : 205-529-2

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only