missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-n-Octyl-D-glucamine, 98%, Thermo Scientific™

$105.74 - $105.74

Chemical Identifiers

CAS 23323-37-7
Molecular Formula C14H31NO5
Molecular Weight (g/mol) 293.404
MDL Number MFCD00134352
InChI Key ZRRNJJURLBXWLL-REWJHTLYSA-N
Synonym n-octyl-d-glucamine, n-octylglucamine, 1-deoxy-1-octylamino-d-glucitol, 2r,3r,4r,5s-6-octylamino hexane-1,2,3,4,5-pentaol, n-n-octyl-d-glucamine, 1-octylamino-1-deoxy-d-glucitol, 1-deoxy-1-n-octylamino-d-glucitol, d-glucitol, 1-deoxy-1-octylamino, 1-deoxy-1-octylamine-d-glucitol, 2r,3r,4r,5s-6-octylamino hexane-1,2,3,4,5-pentol
PubChem CID 90064
IUPAC Name (2R,3R,4R,5S)-6-(octylamino)hexane-1,2,3,4,5-pentol
SMILES CCCCCCCCNCC(C(C(C(CO)O)O)O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6070903
View Documents
Thermo Scientific Chemicals
H6070903
1 g
Each for $105.74
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

23323-37-7
293.404
ZRRNJJURLBXWLL-REWJHTLYSA-N
90064
CCCCCCCCNCC(C(C(C(CO)O)O)O)O
C14H31NO5
MFCD00134352
n-octyl-d-glucamine, n-octylglucamine, 1-deoxy-1-octylamino-d-glucitol, 2r,3r,4r,5s-6-octylamino hexane-1,2,3,4,5-pentaol, n-n-octyl-d-glucamine, 1-octylamino-1-deoxy-d-glucitol, 1-deoxy-1-n-octylamino-d-glucitol, d-glucitol, 1-deoxy-1-octylamino, 1-deoxy-1-octylamine-d-glucitol, 2r,3r,4r,5s-6-octylamino hexane-1,2,3,4,5-pentol
(2R,3R,4R,5S)-6-(octylamino)hexane-1,2,3,4,5-pentol

Specifications

23323-37-7
C14H31NO5
1 g
ZRRNJJURLBXWLL-REWJHTLYSA-N
CCCCCCCCNCC(C(C(C(CO)O)O)O)O
293.404
293.4
N-n-Octyl-D-glucamine
121°C to 124°C
MFCD00134352
n-octyl-d-glucamine, n-octylglucamine, 1-deoxy-1-octylamino-d-glucitol, 2r,3r,4r,5s-6-octylamino hexane-1,2,3,4,5-pentaol, n-n-octyl-d-glucamine, 1-octylamino-1-deoxy-d-glucitol, 1-deoxy-1-n-octylamino-d-glucitol, d-glucitol, 1-deoxy-1-octylamino, 1-deoxy-1-octylamine-d-glucitol, 2r,3r,4r,5s-6-octylamino hexane-1,2,3,4,5-pentol
−15° (c=1 in Methanol)
(2R,3R,4R,5S)-6-(octylamino)hexane-1,2,3,4,5-pentol
90064
98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 245-582-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'