missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Nitrosodibutylamine 97.0+%, TCI America™
Supplier: TCI America N037525ML
Specifications
N-Nitrosodibutylamine | |
237°C | |
MFCD00013892 | |
n-nitrosodibutylamine, dibutylnitrosamine, nitrosodibutylamine, n-nitroso-di-n-butylamine, n-nitrosodi-n-butylamine, 1-butanamine, n-butyl-n-nitroso, ndba, dibutylnitrosoamine, n,n-dibutylnitrosoamine, dbna | |
CCCCN(CCCC)N=O | |
158.245 | |
CHEBI:82356 | |
≥97.0% (GC) |
924-16-3 | |
C8H18N2O | |
25 mL | |
YGJHZCLPZAZIHH-UHFFFAOYSA-N | |
N,N-dibutylnitrous amide | |
13542 | |
158.25 | |
Liquid |
Chemical Identifiers
924-16-3 | |
158.245 | |
YGJHZCLPZAZIHH-UHFFFAOYSA-N | |
13542 | |
N,N-dibutylnitrous amide |
C8H18N2O | |
MFCD00013892 | |
n-nitrosodibutylamine, dibutylnitrosamine, nitrosodibutylamine, n-nitroso-di-n-butylamine, n-nitrosodi-n-butylamine, 1-butanamine, n-butyl-n-nitroso, ndba, dibutylnitrosoamine, n,n-dibutylnitrosoamine, dbna | |
CHEBI:82356 | |
CCCCN(CCCC)N=O |
Safety and Handling
RTECSNumber : EJ4025000
TSCA : Yes