missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Nitrosodiethanolamine 97.0+%, TCI America™
Supplier: TCI America N043825G
Specifications
N-Nitrosodiethanolamine | |
114°C | |
MFCD00015522 | |
Diethanolnitrosamine, NDELA | |
C(CO)N(CCO)N=O | |
134.135 | |
CHEBI:82357 | |
≥97.0% (GC) |
1116-54-7 | |
C4H10N2O3 | |
25 g | |
YFCDLVPYFMHRQZ-UHFFFAOYSA-N | |
N,N-bis(2-hydroxyethyl)nitrous amide | |
14223 | |
134.14 | |
Liquid |
Chemical Identifiers
1116-54-7 | |
134.135 | |
YFCDLVPYFMHRQZ-UHFFFAOYSA-N | |
14223 | |
N,N-bis(2-hydroxyethyl)nitrous amide |
C4H10N2O3 | |
MFCD00015522 | |
Diethanolnitrosamine, NDELA | |
CHEBI:82357 | |
C(CO)N(CCO)N=O |
Safety and Handling
RTECSNumber : KL9550000
TSCA : Yes