missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,O-Bis(trimethylsilyl)acetamide, 95%, Thermo Scientific Chemicals

$61.71 - $186.79

Chemical Identifiers

CAS 10416-59-8
Molecular Formula C8H21NOSi2
Molecular Weight (g/mol) 203.43
MDL Number MFCD00008270
InChI Key SIOVKLKJSOKLIF-UHFFFAOYSA-N
Synonym n,o-bis trimethylsilyl acetamide, trimethylsilyl 1z-n-trimethylsilylethanimidate
PubChem CID 6913588
SMILES CC(O[Si](C)(C)C)=N[Si](C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC156460250
SDS View Documents
Thermo Scientific Chemicals
156460250
25 mL Glass bottle
Each for $61.71
 
AC156461000
SDS View Documents
Thermo Scientific Chemicals
156461000
100 mL Glass bottle
Each for $186.79
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Silylation reagent
Specifications

Chemical Identifiers

10416-59-8
203.43
SIOVKLKJSOKLIF-UHFFFAOYSA-N
6913588
C8H21NOSi2
MFCD00008270
n,o-bis trimethylsilyl acetamide, trimethylsilyl 1z-n-trimethylsilylethanimidate
CC(O[Si](C)(C)C)=N[Si](C)(C)C

Specifications

10416-59-8
100.0
Beige to Brown or Yellow
71°C to 73°C (35.0 mmHg)
Authentic
Glass bottle
1.4165 to 1.4185
MFCD00008270
01,61; 02,30; 03,23; 13,34
n,o-bis trimethylsilyl acetamide, trimethylsilyl 1z-n-trimethylsilylethanimidate
SIOVKLKJSOKLIF-UHFFFAOYSA-N
trimethylsilyl N-(trimethylsilyl)ethanimidate
6913588
95%
N, O-Bis(trimethylsilyl)acetamide
94.0
-8°C
0.8200g/mL
42°C
94% min. (GC)
C8H21NOSi2
CH3C[=NSi(CH3)3]OSi(CH3)3
25 mL
0.82
Solubility in water: decomposes
CC(O[Si](C)(C)C)=N[Si](C)(C)C
203.43
203.43
Crystalline Powder, Crystals or Chunks
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Flammable liquid and vapor.
Reacts violently with water.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 233-892-7

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only