missing translation for 'onlineSavingsMsg'
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits

N,O-Bis(trimethylsilyl)acetamide, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 156460011

Numéro de catalogue. AC156460011

Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Découvrez plus d'offres spéciales

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Silylation reagent
Spécifications

Spécifications

N, O-Bis(trimethylsilyl)acetamide
94.0
0.8200g/mL
42°C
94% min. (GC)
C8H21NOSi2
CH3C[=NSi(CH3)3]OSi(CH3)3
1L
0.82
Solubility in water: decomposes
CC(O[Si](C)(C)C)=N[Si](C)(C)C
203.43
203.43
10416-59-8
100.0
71°C to 73°C (35.0mmHg)
Authentic
Glass bottle
1.4165 to 1.4185
MFCD00008270
01,61; 02,30; 03,23; 13,34
n,o-bis trimethylsilyl acetamide, trimethylsilyl 1z-n-trimethylsilylethanimidate
SIOVKLKJSOKLIF-UHFFFAOYSA-N
trimethylsilyl (1E)-N-trimethylsilylethanimidate
6913588
95%
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Flammable liquid and vapor.
Reacts violently with water.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 233-892-7

FDS Documents

Documents

Offres spéciales

Offres spéciales

RUO – Research Use Only