missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N,O-Bis(trimethylsilyl) acetamide, BSA, EMD-Millipore
Supplier: Technidata 1096490025
Specifications
N, O-Bis(trimethylsilyl) acetamide | |
Glass | |
C8H21NOSi2 | |
MFCD00008270 | |
SIOVKLKJSOKLIF-UHFFFAOYSA-N | |
trimethylsilyl N-(trimethylsilyl)ethanimidate | |
6913588 | |
10 hPa (50°C) |
10416-59-8 | |
25 mL | |
CH3C[NSi(CH3)3]OSi(CH3)3 | |
BSA | |
CC(O[Si](C)(C)C)=N[Si](C)(C)C | |
203.43 | |
203.43 | |
Density: 0.83g/cm3 (20°C) |
Chemical Identifiers
10416-59-8 | |
203.43 | |
SIOVKLKJSOKLIF-UHFFFAOYSA-N | |
6913588 | |
CC(O[Si](C)(C)C)=N[Si](C)(C)C |
C8H21NOSi2 | |
MFCD00008270 | |
BSA | |
trimethylsilyl N-(trimethylsilyl)ethanimidate |
Safety and Handling
Recommended Storage : Store at 15° to 25°C