missing translation for 'onlineSavingsMsg'
Learn More

N,O-Bis(trimethylsilyl)hydroxylamine, 97%

CAS: 22737-37-7 | C6H19NOSi2 | 177.39 g/mol

$72.61 - $239.16

Chemical Identifiers

CAS 22737-37-7
Molecular Formula C6H19NOSi2
Molecular Weight (g/mol) 177.39
MDL Number MFCD00008266
InChI Key ZAEUMMRLGAMWKE-UHFFFAOYSA-N
Synonym n,o-bis trimethylsilyl hydroxylamine, 1,1,1-trimethyl-n-trimethylsilyl oxy silylamine, silanamine, 1,1,1-trimethyl-n-trimethylsilyl oxy, 2,2,5,5-tetramethyl-3-oxa-4-aza-2,5-disilahexane, acmc-209fyl, hydroxylamine, 2tms derivative, n,o-bis-trimethylsilylhydroxylamine, n,o-bis trimethylsilyl hydroxylamin, n,o-bis-trimethylsilyl hydroxylamine
PubChem CID 89815
SMILES C[Si](C)(C)NO[Si](C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC215980010
SDS View Documents
Thermo Scientific Chemicals
215980010
1 g Glass bottle
Each for $72.61
Only null left
 
AC215980050
SDS View Documents
Thermo Scientific Chemicals
215980050
5 g Glass bottle
Each for $239.16
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

22737-37-7
177.39
ZAEUMMRLGAMWKE-UHFFFAOYSA-N
89815
C6H19NOSi2
MFCD00008266
n,o-bis trimethylsilyl hydroxylamine, 1,1,1-trimethyl-n-trimethylsilyl oxy silylamine, silanamine, 1,1,1-trimethyl-n-trimethylsilyl oxy, 2,2,5,5-tetramethyl-3-oxa-4-aza-2,5-disilahexane, acmc-209fyl, hydroxylamine, 2tms derivative, n,o-bis-trimethylsilylhydroxylamine, n,o-bis trimethylsilyl hydroxylamin, n,o-bis-trimethylsilyl hydroxylamine
C[Si](C)(C)NO[Si](C)(C)C

Specifications

22737-37-7
100.0
0.8300g/mL
28°C
96% min. (GC)
C6H19NOSi2
(CH3)3SiONHSi(CH3)3
1 g
0.83
ZAEUMMRLGAMWKE-UHFFFAOYSA-N
2,2,5,5-tetramethyl-3-oxa-4-aza-2,5-disilahexane
89815
97%
N, O-Bis(trimethylsilyl)hydroxylamine, 97%
96.0
Colorless
78.0°C to 80.0°C (100.0 mmHg)
Authentic
Glass bottle
1.4112
MFCD00008266
06,58; 08,523; 09,512; 06,183; 10,142; 12,183; 13,110; 16,128
n,o-bis trimethylsilyl hydroxylamine, 1,1,1-trimethyl-n-trimethylsilyl oxy silylamine, silanamine, 1,1,1-trimethyl-n-trimethylsilyl oxy, 2,2,5,5-tetramethyl-3-oxa-4-aza-2,5-disilahexane, acmc-209fyl, hydroxylamine, 2tms derivative, n,o-bis-trimethylsilylhydroxylamine, n,o-bis trimethylsilyl hydroxylamin, n,o-bis-trimethylsilyl hydroxylamine
C[Si](C)(C)NO[Si](C)(C)C
177.39
177.4
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Causes severe skin burns and eye damage.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin

GHS Signal Word: Danger

EINECSNumber : 245-188-7

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only