Learn More
N,O-Dimethylhydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
$88.95 - $531.42
Chemical Identifiers
CAS | 6638-79-5 |
---|---|
Molecular Formula | C2H8ClNO |
Molecular Weight (g/mol) | 97.54 |
MDL Number | MFCD00012485 |
InChI Key | USZLCYNVCCDPLQ-UHFFFAOYSA-N |
Synonym | n,o-dimethylhydroxylamine hydrochloride, n,o-dimethylhydroxylamine hcl, n-methoxymethylamine hydrochloride, methanamine, n-methoxy-, hydrochloride, o,n-dimethylhydroxylamine hydrochloride, n-methoxymethanamine hydrochloride, o,n-dimethyl-hydroxylamine hydrochloride, methoxy methyl amine hydrochloride, n,o-dimethylhydroxyamine hydrochloride, n,o-dimethylhydroxylaminehydrochloride |
PubChem CID | 81138 |
SMILES | [H+].[Cl-].CNOC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1746906
|
Thermo Scientific Chemicals
A1746906 |
5 g |
Each for $88.95
|
|
||||||
AAA1746914
|
Thermo Scientific Chemicals
A1746914 |
25 g |
Each for $220.97
|
|
||||||
AAA1746922
|
Thermo Scientific Chemicals
A1746922 |
100 g |
Each for $531.42
|
|
||||||
Description
N,O-Dimethylhydroxylamine hydrochloride is used as a reagent in the preparation of 2-acyloxazoles from 2-oxazolemagnesium chloride and N-methoxy-N-methyl-3-oxo-butyramide from diketene. It is also used in amide coupling reactions to form Weinreb amides, which are useful in the Weinreb ketone synthesis. In addition, it is used in the preparation of an inhibitor of the NLS-derived BC peptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN,O-Dimethylhydroxylamine hydrochloride is used as a reagent in the preparation of 2-acyloxazoles from 2-oxazolemagnesium chloride and N-methoxy-N-methyl-3-oxo-butyramide from diketene. It is also used in amide coupling reactions to form Weinreb amides, which are useful in the Weinreb ketone synthesis. In addition, it is used in the preparation of an inhibitor of the NLS-derived BC peptides.
Solubility
Soluble in water, dimethyl sulfoxide and methanol.
Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.
Chemical Identifiers
6638-79-5 | |
97.54 | |
USZLCYNVCCDPLQ-UHFFFAOYSA-N | |
81138 |
C2H8ClNO | |
MFCD00012485 | |
n,o-dimethylhydroxylamine hydrochloride, n,o-dimethylhydroxylamine hcl, n-methoxymethylamine hydrochloride, methanamine, n-methoxy-, hydrochloride, o,n-dimethylhydroxylamine hydrochloride, n-methoxymethanamine hydrochloride, o,n-dimethyl-hydroxylamine hydrochloride, methoxy methyl amine hydrochloride, n,o-dimethylhydroxyamine hydrochloride, n,o-dimethylhydroxylaminehydrochloride | |
[H+].[Cl-].CNOC |
Specifications
6638-79-5 | |
C2H8ClNO | |
5 g | |
Hygroscopic | |
Soluble in water,dimethyl sulfoxide and methanol. | |
[H+].[Cl-].CNOC | |
97.54 | |
97.55 | |
N,O-Dimethylhydroxylamine hydrochloride |
112°C to 115°C | |
MFCD00012485 | |
3650353 | |
n,o-dimethylhydroxylamine hydrochloride, n,o-dimethylhydroxylamine hcl, n-methoxymethylamine hydrochloride, methanamine, n-methoxy-, hydrochloride, o,n-dimethylhydroxylamine hydrochloride, n-methoxymethanamine hydrochloride, o,n-dimethyl-hydroxylamine hydrochloride, methoxy methyl amine hydrochloride, n,o-dimethylhydroxyamine hydrochloride, n,o-dimethylhydroxylaminehydrochloride | |
USZLCYNVCCDPLQ-UHFFFAOYSA-N | |
hydrogen methoxy(methyl)amine chloride | |
81138 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
EINECSNumber : 229-642-1
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only