missing translation for 'onlineSavingsMsg'
Learn More

n-Octadecyl mercaptan, 96%

CAS: 2885-00-9 | C18H38S | 286.56 g/mol

$176.97 - $176.97

Chemical Identifiers

CAS 2885-00-9
Molecular Formula C18H38S
Molecular Weight (g/mol) 286.56
MDL Number MFCD00004886
InChI Key QJAOYSPHSNGHNC-UHFFFAOYSA-N
Synonym octadecanethiol, 1-octadecanethiol, stearyl mercaptan, n-octadecyl mercaptan, octadecyl mercaptan, 1-mercaptooctadecane, octadecanthiol, n-octadecanethiol, octadecanethol, stearylmercaptan
PubChem CID 17905
IUPAC Name octadecane-1-thiol
SMILES CCCCCCCCCCCCCCCCCCS
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC354981000
SDS View Documents
Thermo Scientific Chemicals
354981000
100 g Glass bottle
Each for $176.97
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2885-00-9
286.56
QJAOYSPHSNGHNC-UHFFFAOYSA-N
17905
CCCCCCCCCCCCCCCCCCS
C18H38S
MFCD00004886
octadecanethiol, 1-octadecanethiol, stearyl mercaptan, n-octadecyl mercaptan, octadecyl mercaptan, 1-mercaptooctadecane, octadecanthiol, n-octadecanethiol, octadecanethol, stearylmercaptan
octadecane-1-thiol

Specifications

2885-00-9
100.0
White
185°C
95% min. (GC)
C18H38S
CH3(CH2)16CH2SH
100 g
Solubility in water: insoluble. Other solubilities: soluble in organic solvents
CCCCCCCCCCCCCCCCCCS
286.56
286.56
Crystalline Solid
95.0
25.0°C to 35.0°C
366.0°C
Authentic
Glass bottle
1.4645
MFCD00004886
octadecanethiol, 1-octadecanethiol, stearyl mercaptan, n-octadecyl mercaptan, octadecyl mercaptan, 1-mercaptooctadecane, octadecanthiol, n-octadecanethiol, octadecanethol, stearylmercaptan
QJAOYSPHSNGHNC-UHFFFAOYSA-N
octadecane-1-thiol
17905
96%
n-Octadecyl mercaptan
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause long lasting harmful effects to aquatic life.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protec

GHS Signal Word: Warning

EINECSNumber : 220-744-1

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only