Learn More
n-Octane, 99+%, extra pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 129375000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
n-Octane | |
97 | |
-57.0°C | |
0.7080g/mL | |
13°C | |
99% min. (GC) | |
C8H18 | |
CH3(CH2)6CH3 | |
500 mL | |
01, 159 | |
0.708 | |
n-octane, oktan, oktanen, ottani, oktanen dutch, heptane, methyl, oktan polish, ottani italian, octane, all isomers, unii-x1rv0b2fjv | |
TVMXDCGIABBOFY-UHFFFAOYSA-N | |
octane | |
356 | |
CHEBI:17590 | |
14mbar at 20°C | |
Extra Pure |
111-65-9 | |
100.0 | |
Colorless | |
125.0°C to 127.0°C | |
Authentic | |
Glass Bottle | |
1.3965 to 1.3985 | |
MFCD00009556 | |
1262 | |
0.05% max. | |
15, 6838 | |
Solubility in water: 0.0007g/L (20°C). Other solubilities: soluble in alcohol and acetone, freely miscible with ether, benzene, petroleum, ether and gasoline | |
CCCCCCCC | |
114.23 | |
0.55 mPa.s (20°C) | |
114.23 | |
99+% | |
Liquid |
Chemical Identifiers
111-65-9 | |
114.23 | |
TVMXDCGIABBOFY-UHFFFAOYSA-N | |
356 | |
octane |
C8H18 | |
MFCD00009556 | |
n-octane, oktan, oktanen, ottani, oktanen dutch, heptane, methyl, oktan polish, ottani italian, octane, all isomers, unii-x1rv0b2fjv | |
CHEBI:17590 | |
CCCCCCCC |
Safety and Handling
GHS H Statement
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
May cause drowsiness or dizziness.
May be fatal if swallowed and enters airways.
Highly flammable liquid and vapor.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap
GHS Signal Word: Danger
EINECSNumber : 203-892-1
RUO – Research Use Only