missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
n-Octanoyl Chloride 99.0+%, TCI America™
Supplier: TCI America O0039100ML
Specifications
n-Octanoyl Chloride | |
-63°C | |
195°C | |
MFCD00000766 | |
3265 | |
REEZZSHJLXOIHL-UHFFFAOYSA-N | |
octanoyl chloride | |
8124 | |
≥99.0% (GC) |
111-64-8 | |
Yellow | |
C8H15ClO | |
100 mL | |
capryloyl chloride, caprylyl chloride, n-octanoyl chloride, caprylic acid chloride, octanoic acid chloride, octanoic chloride, octanoylchloride, ccris 5990, caprylic chloride, octanoyl-chloride | |
CCCCCCCC(=O)Cl | |
162.657 | |
162.66 | |
Liquid |
Chemical Identifiers
111-64-8 | |
162.657 | |
REEZZSHJLXOIHL-UHFFFAOYSA-N | |
8124 | |
CCCCCCCC(=O)Cl |
C8H15ClO | |
MFCD00000766 | |
capryloyl chloride, caprylyl chloride, n-octanoyl chloride, caprylic acid chloride, octanoic acid chloride, octanoic chloride, octanoylchloride, ccris 5990, caprylic chloride, octanoyl-chloride | |
octanoyl chloride |
Safety and Handling
EINECSNumber : (2)-0632
RTECSNumber : RH1570000
TSCA : Yes