missing translation for 'onlineSavingsMsg'
Learn More

n-Octanoyl Chloride 99.0+%, TCI America™

Supplier:  TCI America O0039500ML

 View more versions of this product

Catalog No. O0039500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

n-Octanoyl Chloride
-63°C
195°C
MFCD00000766
3265
REEZZSHJLXOIHL-UHFFFAOYSA-N
octanoyl chloride
8124
≥99.0% (GC)
111-64-8
Yellow
C8H15ClO
500 mL
capryloyl chloride, caprylyl chloride, n-octanoyl chloride, caprylic acid chloride, octanoic acid chloride, octanoic chloride, octanoylchloride, ccris 5990, caprylic chloride, octanoyl-chloride
CCCCCCCC(=O)Cl
162.657
162.66
Liquid

Chemical Identifiers

111-64-8
162.657
REEZZSHJLXOIHL-UHFFFAOYSA-N
8124
CCCCCCCC(=O)Cl
C8H15ClO
MFCD00000766
capryloyl chloride, caprylyl chloride, n-octanoyl chloride, caprylic acid chloride, octanoic acid chloride, octanoic chloride, octanoylchloride, ccris 5990, caprylic chloride, octanoyl-chloride
octanoyl chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0632

RTECSNumber : RH1570000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions