missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Octyl-β-D-Glucopyranoside, MP Biomedicals™

n-Octyl-β-D-Glucopyranoside >98%, white powder, non-ionic detergent for membrane protein.

Supplier:  MP Biomedicals Inc 0215005401

 View more versions of this product

Catalog No. ICN15005401

Explore available promotions

Description

Description

  • Non-ionic alkyl-glucoside detergent commonly used in the solubilization of membrane proteins and phospholipids.
  • Used at a molar dependant ratios for the solubilization of phosphatidylcholine bilayers.
  • Used for protein crystallization, as well as for enhancement of oligosaccharide cleavage from glycoproteins using endoglycosidase F.
Specifications

Specifications

n-Octyl-β-D-glucopyranoside
Molecular Biology
Powder
1 g
≥98%
C14H28O6
84118
Soluble in water (10mg/mL-clear colorless solution).
CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O
62852
White
29836-26-8
89°C to 100°C (Literature)
98%
-31 ± 3° (c=5, water)
MFCD00063288
n-Octyl-b-D-glucoside, OGP
HEGSGKPQLMEBJL-RKQHYHRCSA-N
292.37

Chemical Identifiers

29836-26-8
292.37
HEGSGKPQLMEBJL-RKQHYHRCSA-N
62852
C14H28O6
MFCD00063288
n-Octyl-b-D-glucoside, OGP
CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 249-887-8

storageNote1 : 20°C

Recommended Storage : Store at -0°C.

SDS
Documents

Documents

Promotions

Promotions