missing translation for 'onlineSavingsMsg'
Learn More

n-Octyl Ether 96.0+%, TCI America™

$83.89 - $615.14

Chemical Identifiers

CAS 629-82-3
Molecular Formula C16H34O
Molecular Weight (g/mol) 242.447
MDL Number MFCD00009563
InChI Key NKJOXAZJBOMXID-UHFFFAOYSA-N
Synonym Di-n-octyl Ether
PubChem CID 12399
IUPAC Name 1-octoxyoctane
SMILES CCCCCCCCOCCCCCCCC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
O004725ML
View Documents
TCI America
O004725ML
25 mL
Each for $83.89
Only null left
 
O0047100ML
View Documents
TCI America
O0047100ML
100 mL
Each for $212.51
Only null left
 
O0047500ML
View Documents
TCI America
O0047500ML
500 mL
Each for $615.14
Only null left
 
Description

Description

Specifications

Chemical Identifiers

629-82-3
242.447
NKJOXAZJBOMXID-UHFFFAOYSA-N
12399
CCCCCCCCOCCCCCCCC
C16H34O
MFCD00009563
Di-n-octyl Ether
1-octoxyoctane

Specifications

629-82-3
Yellow
C16H34O
25 mL
NKJOXAZJBOMXID-UHFFFAOYSA-N
1-octoxyoctane
12399
≥96.0% (GC)
n-Octyl Ether
-7°C
287°C
MFCD00009563
Di-n-octyl Ether
CCCCCCCCOCCCCCCCC
242.447
242.45
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3074

RTECSNumber : RH8800000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'