missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Octylbenzene, 99%, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals A1213822

 View more versions of this product

Catalog No. AAA1213822


Explore available promotions

Description

Description

n-octylbenzene is used as a solvent to study the self-assembly of derivatives of tetrathiafulvalene (TTF) on graphite and their visualization by scanning tunnelling microscopy (STM). It is also involved in the study the effects and substitution of these compounds. It is also used to prepare charge-transfer complexes with fluoranil and 1,3,5-trinitrobenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-octylbenzene is used as a solvent to study the self-assembly of derivatives of tetrathiafulvalene (TTF) on graphite and their visualization by scanning tunnelling microscopy (STM). It is also involved in the study the effects and substitution of these compounds. It is also used to prepare charge-transfer complexes with fluoranil and 1,3,5-trinitrobenzene.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

n-Octylbenzene
-36°C
264°C to 265°C
Mild
1.484
100 g
n-octylbenzene, 1-phenyloctane, benzene, octyl, phenyloctane, octane, 1-phenyl, octyl-benzene, 1-octylbenzene, octyl benzene, 4-octylbenzene, acmc-1cplg
CDKDZKXSXLNROY-UHFFFAOYSA-N
octylbenzene
16607
99%
2189-60-8
0.854
107°C (224°F)
C14H22
MFCD00009564
1906253
Immiscible with water.
CCCCCCCCC1=CC=CC=C1
190.33
190.33

Chemical Identifiers

2189-60-8
190.33
CDKDZKXSXLNROY-UHFFFAOYSA-N
16607
CCCCCCCCC1=CC=CC=C1
C14H22
MFCD00009564
n-octylbenzene, 1-phenyloctane, benzene, octyl, phenyloctane, octane, 1-phenyl, octyl-benzene, 1-octylbenzene, octyl benzene, 4-octylbenzene, acmc-1cplg
octylbenzene
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 218-582-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only