missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
n-Pentane For gas chromatography ECD and FID SupraSolv™, MilliporeSigma™
For gas chromatography ECD and FID SupraSolv™
$460.42 - $959.97
Chemical Identifiers
CAS | 109-66-0 |
---|---|
Molecular Formula | C5H12 |
Molecular Weight (g/mol) | 72.15 |
MDL Number | MFCD00009498 |
InChI Key | OFBQJSOFQDEBGM-UHFFFAOYSA-N |
Synonym | n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish |
PubChem CID | 8003 |
ChEBI | CHEBI:37830 |
IUPAC Name | pentane |
SMILES | CCCCC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
M1008821000
|
Technidata
1008821000 |
1 L | Glass Bottle |
Each for $460.42
|
|
||||
M1008822500
|
Technidata
1008822500 |
2.5 L | Glass Bottle |
Each for $959.97
|
|
||||
Chemical Identifiers
109-66-0 | |
72.15 | |
OFBQJSOFQDEBGM-UHFFFAOYSA-N | |
8003 | |
pentane |
C5H12 | |
MFCD00009498 | |
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish | |
CHEBI:37830 | |
CCCCC |
Specifications
-130°C | |
Colorless | |
36°C | |
48°C (54°F) | |
Benzine-like | |
Liquid | |
MFCD00009498 | |
1265 | |
Soluble in water: 0.4g/L at 16°C | |
CCCCC | |
72.15 | |
CHEBI:37830 | |
n-Pentane |
109-66-0 | |
0.63g/cm3 at 20°C | |
1 L | |
285°C (545°F) | |
Glass Bottle | |
C5H12 | |
570 hPa (20°C) | |
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish | |
OFBQJSOFQDEBGM-UHFFFAOYSA-N | |
pentane | |
8003 | |
72.15 |
Safety and Handling
H304: May be fatal if swallowed and enters airways.
H336 May cause drowsiness and dizziness.
H225: Highly flammable liquid and vapour.
Recommended Storage : 5°C to 30°C (41°F to 86°F); Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition.