Learn More
N-Phenylbis(trifluoromethanesulfonimide), 97%, Thermo Scientific Chemicals
$126.57 - $426.33
Chemical Identifiers
CAS | 37595-74-7 |
---|---|
Molecular Formula | C8H5F6NO4S2 |
Molecular Weight (g/mol) | 357.25 |
MDL Number | MFCD00000404 |
InChI Key | DIOHEXPTUTVCNX-UHFFFAOYSA-N |
Synonym | n,n-bis trifluoromethylsulfonyl aniline, phenyl triflimide, n-phenyl-bis trifluoromethanesulfonimide, n-phenyltrifluoromethanesulfonimide, n,n-bis-trifluoromethanesulfonyl aniline, 1,1,1-trifluoro-n-phenyl-n-trifluoromethylsulfonyl methanesulfonamide, n-phenylbis trifluoromethanesulfonimide, n-phenylbis trifluoromethanesulphonimide, 1,1,1-trifluoro-n-phenyl-n-trifluoromethyl sulfonyl methanesulfonamide, n,n-bis trifluoromethanesulfonyl aniline |
PubChem CID | 142176 |
IUPAC Name | 1,1,1-trifluoro-N-phenyl-N-(trifluoromethylsulfonyl)methanesulfonamide |
SMILES | C1=CC=C(C=C1)N(S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC374790050
|
Thermo Scientific Chemicals
374790050 |
5 g | Glass bottle |
Each for $126.57
|
|
||||
AC374790250
|
Thermo Scientific Chemicals
374790250 |
25 g | Glass bottle |
Each for $426.33
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagentChemical Identifiers
37595-74-7 | |
357.25 | |
DIOHEXPTUTVCNX-UHFFFAOYSA-N | |
142176 | |
C1=CC=C(C=C1)N(S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F |
C8H5F6NO4S2 | |
MFCD00000404 | |
n,n-bis trifluoromethylsulfonyl aniline, phenyl triflimide, n-phenyl-bis trifluoromethanesulfonimide, n-phenyltrifluoromethanesulfonimide, n,n-bis-trifluoromethanesulfonyl aniline, 1,1,1-trifluoro-n-phenyl-n-trifluoromethylsulfonyl methanesulfonamide, n-phenylbis trifluoromethanesulfonimide, n-phenylbis trifluoromethanesulphonimide, 1,1,1-trifluoro-n-phenyl-n-trifluoromethyl sulfonyl methanesulfonamide, n,n-bis trifluoromethanesulfonyl aniline | |
1,1,1-trifluoro-N-phenyl-N-(trifluoromethylsulfonyl)methanesulfonamide |
Specifications
37595-74-7 | |
100.0 | |
White or Colorless | |
96% min. (HPLC) | |
C8H5F6NO4S2 | |
MFCD00000404 | |
15,7427 | |
DIOHEXPTUTVCNX-UHFFFAOYSA-N | |
1,1,1-trifluoro-N-phenyl-N-(trifluoromethylsulfonyl)methanesulfonamide | |
142176 | |
97% | |
N-Phenylbis(trifluoromethanesulfonimide) |
96.0 | |
99.0°C to 105.0°C | |
Authentic | |
Glass bottle | |
C6H5N(SO2CF3)2 | |
5 g | |
n,n-bis trifluoromethylsulfonyl aniline, phenyl triflimide, n-phenyl-bis trifluoromethanesulfonimide, n-phenyltrifluoromethanesulfonimide, n,n-bis-trifluoromethanesulfonyl aniline, 1,1,1-trifluoro-n-phenyl-n-trifluoromethylsulfonyl methanesulfonamide, n-phenylbis trifluoromethanesulfonimide, n-phenylbis trifluoromethanesulphonimide, 1,1,1-trifluoro-n-phenyl-n-trifluoromethyl sulfonyl methanesulfonamide, n,n-bis trifluoromethanesulfonyl aniline | |
C1=CC=C(C=C1)N(S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F | |
357.25 | |
357.25 | |
Crystalline Powder or Crystals |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
RUO – Research Use Only