missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Phenylethanolamine, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2213136

 View more versions of this product

Catalog No. AAB2213136


Explore available promotions

Description

Description

N-Phenylethanolamine is used as an intermediate for dyes and other organic compounds. Further, it is used as a substrate for human olfactory uridine 5'-diphospho-glucuronosyltransferase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Phenylethanolamine is used as an intermediate for dyes and other organic compounds. Further, it is used as a substrate for human olfactory uridine 5′-diphospho-glucuronosyltransferase.

Solubility
Slightly miscible with water.

Notes
Incompatible with acids, acid anhydrides, acid chlorides and chloroformates.
Specifications

Specifications

N-Phenylethanolamine
-30°C
10.2
153°C (307°F)
C8H11NO
MFCD00002832
UN2810
2-phenylamino ethanol, n-phenylethanolamine, n-2-hydroxyethyl aniline, ethanol, 2-phenylamino, 2-phenylamino ethan-1-ol, ethanol, 2-anilino, beta-anilinoethanol, aniline, n-2-hydroxyethyl, n-2-hydroxyethyl phenylamine, emery 5700
MWGATWIBSKHFMR-UHFFFAOYSA-N
2-anilinoethanol
31235
98%
122-98-5
1.087
284°C to 286°C
Amine-like
1.579
500 g
774672
Slightly miscible with water.
C1=CC=C(C=C1)NCCO
137.182
137.18

Chemical Identifiers

122-98-5
137.182
MWGATWIBSKHFMR-UHFFFAOYSA-N
31235
C1=CC=C(C=C1)NCCO
C8H11NO
MFCD00002832
2-phenylamino ethanol, n-phenylethanolamine, n-2-hydroxyethyl aniline, ethanol, 2-phenylamino, 2-phenylamino ethan-1-ol, ethanol, 2-anilino, beta-anilinoethanol, aniline, n-2-hydroxyethyl, n-2-hydroxyethyl phenylamine, emery 5700
2-anilinoethanol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H310-H351-H373-H319-H317
Fatal in contact with skin.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Causes serious eye irritation.
May cause an allergic skin reaction.

P264b-P280-P302+P352-P305+P351+P338-P312-P361-P363-P501c

H311-H319

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

EINECSNumber : 204-588-1

RTECSNumber : KJ7175000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only