Learn More
N-Phenylmaleimide, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 130711000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| N-Phenylmaleimide | |
| 941-69-5 | |
| 100.0 | |
| Authentic | |
| Glass bottle | |
| MFCD00005502 | |
| 21, 400 | |
| 15, 7410 | |
| HIDBROSJWZYGSZ-UHFFFAOYSA-N | |
| 1-phenylpyrrole-2,5-dione | |
| 13662 | |
| 97% |
| 97% | |
| 96.0 | |
| 162°C to 163°C (12.0mmHg) | |
| 96% min. (GC) | |
| C10H7NO2 | |
| 100g | |
| 01,845; 13,317 | |
| n-phenylmaleimide, maleanil, maleinanil, maleimidobenzene, 1h-pyrrole-2,5-dione, 1-phenyl, 1-phenyl-1h-pyrrole-2,5-dione, maleimide, n-phenyl, 1-phenyl-2,5-dihydro-1h-pyrrole-2,5-dione, unii-9u9kt462vw, n-fenylimid kyseliny maleinove | |
| C1=CC=C(C=C1)N2C(=O)C=CC2=O | |
| 173.17 | |
| 173.17 |
Chemical Identifiers
| 941-69-5 | |
| 173.17 | |
| HIDBROSJWZYGSZ-UHFFFAOYSA-N | |
| 13662 | |
| C1=CC=C(C=C1)N2C(=O)C=CC2=O |
| C10H7NO2 | |
| MFCD00005502 | |
| n-phenylmaleimide, maleanil, maleinanil, maleimidobenzene, 1h-pyrrole-2,5-dione, 1-phenyl, 1-phenyl-1h-pyrrole-2,5-dione, maleimide, n-phenyl, 1-phenyl-2,5-dihydro-1h-pyrrole-2,5-dione, unii-9u9kt462vw, n-fenylimid kyseliny maleinove | |
| 1-phenylpyrrole-2,5-dione |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Toxic if swallowed.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Do not eat,drink or smoke when using this product.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy
GHS Signal Word: Danger
EINECSNumber : 213-382-0
RTECSNumber : ON5950000
TSCA : TSCA
RUO â Research Use Only