missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Phenylsuccinimide 98.0+%, TCI America™
$164.03 - $484.65
Chemical Identifiers
| CAS | 83-25-0 |
|---|---|
| Molecular Formula | C10H9NO2 |
| Molecular Weight (g/mol) | 175.187 |
| MDL Number | MFCD00022587 |
| InChI Key | ZTUKZULGOCFJET-UHFFFAOYSA-N |
| PubChem CID | 66519 |
| IUPAC Name | 1-phenylpyrrolidine-2,5-dione |
| SMILES | C1CC(=O)N(C1=O)C2=CC=CC=C2 |
Chemical Identifiers
| 83-25-0 | |
| 175.187 | |
| ZTUKZULGOCFJET-UHFFFAOYSA-N | |
| 1-phenylpyrrolidine-2,5-dione |
| C10H9NO2 | |
| MFCD00022587 | |
| 66519 | |
| C1CC(=O)N(C1=O)C2=CC=CC=C2 |
Specifications
| 83-25-0 | |
| White | |
| MFCD00022587 | |
| ZTUKZULGOCFJET-UHFFFAOYSA-N | |
| 1-phenylpyrrolidine-2,5-dione | |
| 66519 | |
| ≥98.0% (GC) | |
| N-Phenylsuccinimide |
| 155°C | |
| C10H9NO2 | |
| 5 g | |
| C1CC(=O)N(C1=O)C2=CC=CC=C2 | |
| 175.187 | |
| 175.19 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator