missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Phenylthiourea, 97%, Thermo Scientific Chemicals

$78.55 - $700.42

Chemical Identifiers

CAS 103-85-5
Molecular Formula C7H8N2S
Molecular Weight (g/mol) 152.22
MDL Number MFCD00004933
InChI Key FULZLIGZKMKICU-UHFFFAOYSA-N
Synonym n-phenylthiourea, 1-phenylthiourea, 1-phenyl-2-thiourea, phenylthiocarbamide, thiourea, phenyl, phenyl-2-thiourea, fenylthiomocovina, urea, 1-phenyl-2-thio, rcra waste number p093, phenyl-thiourea
PubChem CID 676454
ChEBI CHEBI:46261
IUPAC Name phenylthiourea
SMILES NC(=S)NC1=CC=CC=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0669009
View Documents
Thermo Scientific Chemicals
L0669009
10 g
Each for $78.55
 
AAL0669018
View Documents
Thermo Scientific Chemicals
L0669018
50 g
Each for $239.96
 
AAL0669030
View Documents
Thermo Scientific Chemicals
L0669030
250 g
Each for $700.42
 
Description

Description

N-phenylthiourea is used in paternity testing prior to the advent of DNA testing. It inhibits melanogenesis and is used to grow transparent fish. It is also used as a repellent for rat, rabbits, weasels and also in medical genetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-phenylthiourea is used in paternity testing prior to the advent of DNA testing. It inhibits melanogenesis and is used to grow transparent fish. It is also used as a repellent for rat, rabbits, weasels and also in medical genetics.

Solubility
Soluble in hot water and alcohol.

Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.
Specifications

Chemical Identifiers

103-85-5
152.22
FULZLIGZKMKICU-UHFFFAOYSA-N
676454
phenylthiourea
C7H8N2S
MFCD00004933
n-phenylthiourea, 1-phenylthiourea, 1-phenyl-2-thiourea, phenylthiocarbamide, thiourea, phenyl, phenyl-2-thiourea, fenylthiomocovina, urea, 1-phenyl-2-thio, rcra waste number p093, phenyl-thiourea
CHEBI:46261
NC(=S)NC1=CC=CC=C1

Specifications

103-85-5
Odorless
MFCD00004933
UN2811
14,7315
Soluble in hot water and alcohol.
NC(=S)NC1=CC=CC=C1
152.22
CHEBI:46261
97%
149°C to 153°C
C7H8N2S
10 g
907309
n-phenylthiourea, 1-phenylthiourea, 1-phenyl-2-thiourea, phenylthiocarbamide, thiourea, phenyl, phenyl-2-thiourea, fenylthiomocovina, urea, 1-phenyl-2-thio, rcra waste number p093, phenyl-thiourea
FULZLIGZKMKICU-UHFFFAOYSA-N
phenylthiourea
676454
152.21
N-Phenylthiourea
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300
Fatal if swallowed.

P261-P264b-P270-P272-P280g-P301+P310-P302+P352-P330-P333+P313-P363-P501c

H300-H317

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 203-151-2

RTECSNumber : YU1400000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only