missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Phenylthiourea 98.0+%, TCI America™

Supplier:  TCI America P0237500G

 View more versions of this product

Catalog No. P0237500G


Explore available promotions

Description

Description

Specifications

Specifications

N-Phenylthiourea
153°C
C7H8N2S
500 g
n-phenylthiourea, 1-phenylthiourea, 1-phenyl-2-thiourea, phenylthiocarbamide, thiourea, phenyl, phenyl-2-thiourea, fenylthiomocovina, urea, 1-phenyl-2-thio, rcra waste number p093, phenyl-thiourea
NC(=S)NC1=CC=CC=C1
152.22
CHEBI:46261
≥98.0% (HPLC,N)
103-85-5
White
MFCD00004933
2811
FULZLIGZKMKICU-UHFFFAOYSA-N
phenylthiourea
676454
152.22
Crystalline Powder

Chemical Identifiers

103-85-5
152.22
FULZLIGZKMKICU-UHFFFAOYSA-N
676454
phenylthiourea
C7H8N2S
MFCD00004933
n-phenylthiourea, 1-phenylthiourea, 1-phenyl-2-thiourea, phenylthiocarbamide, thiourea, phenyl, phenyl-2-thiourea, fenylthiomocovina, urea, 1-phenyl-2-thio, rcra waste number p093, phenyl-thiourea
CHEBI:46261
NC(=S)NC1=CC=CC=C1
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-3230

RTECSNumber : YU1400000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions