missing translation for 'onlineSavingsMsg'
Learn More

N-Pivaloyl-o-toluidine, 99%, Thermo Scientific™

Chemical Identifiers

CAS 61495-04-3
Molecular Formula C12H17NO
Molecular Weight (g/mol) 191.27
MDL Number MFCD00075432
InChI Key CSGRQLUGMVFNON-UHFFFAOYSA-N
Synonym n-o-tolyl pivalamide, 2,2,2'-trimethylpropionanilide, 2,2-dimethyl-n-2-methylphenyl propanamide, n-o-tolylpivalamide, acmc-1b8p7, n-2-methylphenyl pivalamide, 2,2-dimethyl-n-o-tolyl-propionamide, 2,2-dimethyl-n-2-methyl phenyl propanamide, n-2-methyl phenyl-2,2-dimethylpropanamide
PubChem CID 2734005
IUPAC Name 2,2-dimethyl-N-(2-methylphenyl)propanamide
SMILES CC1=CC=CC=C1NC(=O)C(C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC346730050
SDS View Documents
Thermo Scientific Chemicals
346730050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC346730250
SDS View Documents
Thermo Scientific Chemicals
346730250
25g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

61495-04-3
191.27
CSGRQLUGMVFNON-UHFFFAOYSA-N
2734005
CC1=CC=CC=C1NC(=O)C(C)(C)C
C12H17NO
MFCD00075432
n-o-tolyl pivalamide, 2,2,2'-trimethylpropionanilide, 2,2-dimethyl-n-2-methylphenyl propanamide, n-o-tolylpivalamide, acmc-1b8p7, n-2-methylphenyl pivalamide, 2,2-dimethyl-n-o-tolyl-propionamide, 2,2-dimethyl-n-2-methyl phenyl propanamide, n-2-methyl phenyl-2,2-dimethylpropanamide
2,2-dimethyl-N-(2-methylphenyl)propanamide

Specifications

61495-04-3
100.0
98.5% min. (GC)
C12H17NO
5g
CSGRQLUGMVFNON-UHFFFAOYSA-N
2,2-dimethyl-N-(2-methylphenyl)propanamide
2734005
99%
98.5
Authentic
Glass bottle
MFCD00075432
n-o-tolyl pivalamide, 2,2,2'-trimethylpropionanilide, 2,2-dimethyl-n-2-methylphenyl propanamide, n-o-tolylpivalamide, acmc-1b8p7, n-2-methylphenyl pivalamide, 2,2-dimethyl-n-o-tolyl-propionamide, 2,2-dimethyl-n-2-methyl phenyl propanamide, n-2-methyl phenyl-2,2-dimethylpropanamide
CC1=CC=CC=C1NC(=O)C(C)(C)C
191.27
191.27
N-Pivaloyl-o-toluidine, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only