missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Propyl Alcohol, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.5%, Honeywell

Supplier:  Honeywell Inc LP3224

 View more versions of this product

Catalog No. 6000108


Explore available promotions

Description

Description

Specifications

Specifications

n-Propyl Alcohol
71-23-8
0.8053 g/cm3
4 x 4 L
Glass Bottle
C3H8O
UN1274
1098242
Miscible in Water at 20°C
CCCO
60.10
CHEBI:28831
>99.5%
ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.5%
100.0000%
97.2°C
23°C
by GC Analysis
MFCD00002941
0.2% max.
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
BDERNNFJNOPAEC-UHFFFAOYSA-N
propan-1-ol
1031
60.09
ACS Reagent

Chemical Identifiers

71-23-8
60.10
BDERNNFJNOPAEC-UHFFFAOYSA-N
1031
propan-1-ol
C3H8O
MFCD00002941
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
CHEBI:28831
CCCO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 547 days from date of manufacture

DOTInformation : Transport Hazard Class: 3; Packing Group:3: III; Proper Shipping Name: n-Propanol

SDS
Documents

Documents

Promotions

Promotions