missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate 98.0+%, TCI America™

Supplier:  TCI America S0853100MG

 View more versions of this product

Catalog No. S0853100MG


Explore available promotions

Description

Description

Specifications

Specifications

N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate
177°C
C16H18N2O6
100 mg
JJAHTWIKCUJRDK-UHFFFAOYSA-N
(2,5-dioxopyrrolidin-1-yl) 4-[(2,5-dioxopyrrol-1-yl)methyl]cyclohexane-1-carboxylate
125175
334.33
Crystalline Powder
64987-85-5
White
MFCD00009634
smcc, 2,5-dioxopyrrolidin-1-yl 4-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl methyl cyclohexanecarboxylate, n-succinimidyl 4-n-maleimidomethyl cyclohexane-1-carboxylate, trans-4-maleimidomethyl cyclohexanecarboxylic acid-nhs, unii-b357p1g1if, n-succinimidyl 4-maleimidomethyl cyclohexane-1-carboxylate, n-4-carboxycyclohexylmethyl maleimide n-hydroxysuccinimide ester, trans-2,5-dioxopyrrolidin-1-yl 4-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl methyl cyclohexanecarboxylate, n-hydroxysuccinimidyl 4-n-maleimidomethylcyclohexane-1-carboxylate, succinimidyl 4-n-maleimidomethyl cyclohexane-1-carboxylate
C1CC(CCC1CN2C(=O)C=CC2=O)C(=O)ON3C(=O)CCC3=O
334.328
CHEBI:63174
≥98.0% (HPLC)

Chemical Identifiers

64987-85-5
334.328
JJAHTWIKCUJRDK-UHFFFAOYSA-N
125175
(2,5-dioxopyrrolidin-1-yl) 4-[(2,5-dioxopyrrol-1-yl)methyl]cyclohexane-1-carboxylate
C16H18N2O6
MFCD00009634
smcc, 2,5-dioxopyrrolidin-1-yl 4-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl methyl cyclohexanecarboxylate, n-succinimidyl 4-n-maleimidomethyl cyclohexane-1-carboxylate, trans-4-maleimidomethyl cyclohexanecarboxylic acid-nhs, unii-b357p1g1if, n-succinimidyl 4-maleimidomethyl cyclohexane-1-carboxylate, n-4-carboxycyclohexylmethyl maleimide n-hydroxysuccinimide ester, trans-2,5-dioxopyrrolidin-1-yl 4-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl methyl cyclohexanecarboxylate, n-hydroxysuccinimidyl 4-n-maleimidomethylcyclohexane-1-carboxylate, succinimidyl 4-n-maleimidomethyl cyclohexane-1-carboxylate
CHEBI:63174
C1CC(CCC1CN2C(=O)C=CC2=O)C(=O)ON3C(=O)CCC3=O
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions