missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate 98.0+%, TCI America™
Supplier: TCI America S08531G
Specifications
N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate | |
177°C | |
C16H18N2O6 | |
1 g | |
JJAHTWIKCUJRDK-UHFFFAOYSA-N | |
(2,5-dioxopyrrolidin-1-yl) 4-[(2,5-dioxopyrrol-1-yl)methyl]cyclohexane-1-carboxylate | |
125175 | |
334.33 | |
Crystalline Powder |
64987-85-5 | |
White | |
MFCD00009634 | |
smcc, 2,5-dioxopyrrolidin-1-yl 4-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl methyl cyclohexanecarboxylate, n-succinimidyl 4-n-maleimidomethyl cyclohexane-1-carboxylate, trans-4-maleimidomethyl cyclohexanecarboxylic acid-nhs, unii-b357p1g1if, n-succinimidyl 4-maleimidomethyl cyclohexane-1-carboxylate, n-4-carboxycyclohexylmethyl maleimide n-hydroxysuccinimide ester, trans-2,5-dioxopyrrolidin-1-yl 4-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl methyl cyclohexanecarboxylate, n-hydroxysuccinimidyl 4-n-maleimidomethylcyclohexane-1-carboxylate, succinimidyl 4-n-maleimidomethyl cyclohexane-1-carboxylate | |
C1CC(CCC1CN2C(=O)C=CC2=O)C(=O)ON3C(=O)CCC3=O | |
334.328 | |
CHEBI:63174 | |
≥98.0% (HPLC) |
Chemical Identifiers
64987-85-5 | |
334.328 | |
JJAHTWIKCUJRDK-UHFFFAOYSA-N | |
125175 | |
(2,5-dioxopyrrolidin-1-yl) 4-[(2,5-dioxopyrrol-1-yl)methyl]cyclohexane-1-carboxylate |
C16H18N2O6 | |
MFCD00009634 | |
smcc, 2,5-dioxopyrrolidin-1-yl 4-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl methyl cyclohexanecarboxylate, n-succinimidyl 4-n-maleimidomethyl cyclohexane-1-carboxylate, trans-4-maleimidomethyl cyclohexanecarboxylic acid-nhs, unii-b357p1g1if, n-succinimidyl 4-maleimidomethyl cyclohexane-1-carboxylate, n-4-carboxycyclohexylmethyl maleimide n-hydroxysuccinimide ester, trans-2,5-dioxopyrrolidin-1-yl 4-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl methyl cyclohexanecarboxylate, n-hydroxysuccinimidyl 4-n-maleimidomethylcyclohexane-1-carboxylate, succinimidyl 4-n-maleimidomethyl cyclohexane-1-carboxylate | |
CHEBI:63174 | |
C1CC(CCC1CN2C(=O)C=CC2=O)C(=O)ON3C(=O)CCC3=O |
Safety and Handling
TSCA : No
Recommended Storage : Freezer