missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Succinimidyl Acetate 98.0+%, TCI America™
$123.03 - $369.08
Chemical Identifiers
| CAS | 14464-29-0 |
|---|---|
| Molecular Formula | C6H7NO4 |
| Molecular Weight (g/mol) | 157.125 |
| MDL Number | MFCD00037916 |
| InChI Key | SIFCHNIAAPMMKG-UHFFFAOYSA-N |
| Synonym | Acetic Acid N-Succinimidyl Ester, N-Acetoxysuccinimide |
| PubChem CID | 84460 |
| IUPAC Name | (2,5-dioxopyrrolidin-1-yl) acetate |
| SMILES | CC(=O)ON1C(=O)CCC1=O |
Chemical Identifiers
| 14464-29-0 | |
| 157.125 | |
| SIFCHNIAAPMMKG-UHFFFAOYSA-N | |
| 84460 | |
| CC(=O)ON1C(=O)CCC1=O |
| C6H7NO4 | |
| MFCD00037916 | |
| Acetic Acid N-Succinimidyl Ester, N-Acetoxysuccinimide | |
| (2,5-dioxopyrrolidin-1-yl) acetate |
Specifications
| 14464-29-0 | |
| White | |
| MFCD00037916 | |
| Acetic Acid N-Succinimidyl Ester, N-Acetoxysuccinimide | |
| CC(=O)ON1C(=O)CCC1=O | |
| 157.125 | |
| 157.13 | |
| Crystalline Powder |
| 134°C | |
| C6H7NO4 | |
| 5 g | |
| SIFCHNIAAPMMKG-UHFFFAOYSA-N | |
| (2,5-dioxopyrrolidin-1-yl) acetate | |
| 84460 | |
| ≥98.0% (GC) | |
| N-Succinimidyl Acetate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator