missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Succinimidyl Tetradecanoate 95.0+%, TCI America™
$318.75 - $958.13
Chemical Identifiers
| CAS | 69888-86-4 |
|---|---|
| Molecular Formula | C18H31NO4 |
| Molecular Weight (g/mol) | 325.449 |
| MDL Number | MFCD00522592 |
| InChI Key | NZEDHZOVUDEBGW-UHFFFAOYSA-N |
| Synonym | Tetradecanoic Acid N-Succinimidyl Ester, N-Succinimidyl Myristate, Myristic Acid N-Succinimidyl Ester, NHS-Myristate |
| PubChem CID | 4229289 |
| IUPAC Name | (2,5-dioxopyrrolidin-1-yl) tetradecanoate |
| SMILES | CCCCCCCCCCCCCC(=O)ON1C(=O)CCC1=O |
Chemical Identifiers
| 69888-86-4 | |
| 325.449 | |
| NZEDHZOVUDEBGW-UHFFFAOYSA-N | |
| 4229289 | |
| CCCCCCCCCCCCCC(=O)ON1C(=O)CCC1=O |
| C18H31NO4 | |
| MFCD00522592 | |
| Tetradecanoic Acid N-Succinimidyl Ester, N-Succinimidyl Myristate, Myristic Acid N-Succinimidyl Ester, NHS-Myristate | |
| (2,5-dioxopyrrolidin-1-yl) tetradecanoate |
Specifications
| 69888-86-4 | |
| White | |
| MFCD00522592 | |
| Tetradecanoic Acid N-Succinimidyl Ester, N-Succinimidyl Myristate, Myristic Acid N-Succinimidyl Ester, NHS-Myristate | |
| CCCCCCCCCCCCCC(=O)ON1C(=O)CCC1=O | |
| 325.449 | |
| 325.45 | |
| Crystalline Powder |
| 84°C | |
| C18H31NO4 | |
| 1 g | |
| NZEDHZOVUDEBGW-UHFFFAOYSA-N | |
| (2,5-dioxopyrrolidin-1-yl) tetradecanoate | |
| 4229289 | |
| ≥95.0% (HPLC,N) | |
| N-Succinimidyl Tetradecanoate |
Safety and Handling
TSCA : No
Recommended Storage : Freezer (-20 °C)