missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)iminodiacetic Acid 98.0+%, TCI America™

$61.51 - $208.78

Chemical Identifiers

CAS 56074-20-5
Molecular Formula C9H15NO6
Molecular Weight (g/mol) 233.22
MDL Number MFCD00800351
InChI Key QPGIJQUTGABQLQ-UHFFFAOYSA-N
Synonym n-boc-iminodiacetic acid, boc-dl-ida-oh, 2,2'-tert-butoxycarbonyl azanediyl diacetic acid, tert-butoxycarbonyl carboxymethyl amino acetic acid, n-tert-butoxycarbonyl iminodiacetic acid, glycine, n-carboxymethyl-n-1,1-dimethylethoxy carbonyl, 2-carboxymethyl-2-methylpropan-2-yl oxycarbonyl amino acetic acid, tert-butoxycarbonylimino bisacetic acid, n-t-butyloxycarbonyl iminodiacetic acid, n-tert-butoxycarbonyl-iminodiacetic acid
PubChem CID 2755971
IUPAC Name 2-[carboxymethyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]acetic acid
SMILES CC(C)(C)OC(=O)N(CC(=O)O)CC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B43685G
View Documents
TCI America
B43685G
5 g
Each for $61.51
Only null left
 
B436825G
View Documents
TCI America
B436825G
25 g
Each for $208.78
Only null left
 
Description

Description

Specifications

Chemical Identifiers

56074-20-5
233.22
QPGIJQUTGABQLQ-UHFFFAOYSA-N
2755971
CC(C)(C)OC(=O)N(CC(=O)O)CC(=O)O
C9H15NO6
MFCD00800351
n-boc-iminodiacetic acid, boc-dl-ida-oh, 2,2'-tert-butoxycarbonyl azanediyl diacetic acid, tert-butoxycarbonyl carboxymethyl amino acetic acid, n-tert-butoxycarbonyl iminodiacetic acid, glycine, n-carboxymethyl-n-1,1-dimethylethoxy carbonyl, 2-carboxymethyl-2-methylpropan-2-yl oxycarbonyl amino acetic acid, tert-butoxycarbonylimino bisacetic acid, n-t-butyloxycarbonyl iminodiacetic acid, n-tert-butoxycarbonyl-iminodiacetic acid
2-[carboxymethyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]acetic acid

Specifications

126°C
White
MFCD00800351
QPGIJQUTGABQLQ-UHFFFAOYSA-N
2-[carboxymethyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]acetic acid
233.22
233.22
Crystalline Powder
56074-20-5
C9H15NO6
n-boc-iminodiacetic acid, boc-dl-ida-oh, 2,2'-tert-butoxycarbonyl azanediyl diacetic acid, tert-butoxycarbonyl carboxymethyl amino acetic acid, n-tert-butoxycarbonyl iminodiacetic acid, glycine, n-carboxymethyl-n-1,1-dimethylethoxy carbonyl, 2-carboxymethyl-2-methylpropan-2-yl oxycarbonyl amino acetic acid, tert-butoxycarbonylimino bisacetic acid, n-t-butyloxycarbonyl iminodiacetic acid, n-tert-butoxycarbonyl-iminodiacetic acid
CC(C)(C)OC(=O)N(CC(=O)O)CC(=O)O
5 g
2755971
≥98.0% (HPLC,T)
N-(tert-Butoxycarbonyl)iminodiacetic Acid
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
missing translation for 'documents'

missing translation for 'documents'