missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-(tert-Butoxycarbonyl)-L-tert-leucine 98.0+%, TCI America™
Supplier: TCI America B375425G
Specifications
119°C | |
62965-35-9 | |
C11H21NO4 | |
n-boc-l-tert-leucine, boc-tle-oh, boc-l-tert-leucine, s-n-tert-butoxycarbonyl-tert-leucine, s-2-tert-butoxycarbonyl amino-3,3-dimethylbutanoic acid, boc-l-alpha-t-butylglycine, boc-alpha-t-butylglycine, boc-tbu-gly-oh, boc-l-tle-oh, n-tert-butoxycarbonyl-l-tert-leucine | |
CC(C)(C)OC(=O)N[C@H](C(O)=O)C(C)(C)C | |
25 g | |
2734668 | |
≥98.0% (HPLC,T) |
N-(tert-Butoxycarbonyl)-L-tert-leucine | |
White | |
MFCD00065574 | |
LRFZIPCTFBPFLX-JLDDOWRYNA-N | |
(2S)-2-{[(tert-butoxy)carbonyl]amino}-3,3-dimethylbutanoic acid | |
231.29 | |
231.29 | |
Crystalline Powder |
Chemical Identifiers
62965-35-9 | |
231.29 | |
LRFZIPCTFBPFLX-JLDDOWRYNA-N | |
2734668 | |
CC(C)(C)OC(=O)N[C@H](C(O)=O)C(C)(C)C |
C11H21NO4 | |
MFCD00065574 | |
n-boc-l-tert-leucine, boc-tle-oh, boc-l-tert-leucine, s-n-tert-butoxycarbonyl-tert-leucine, s-2-tert-butoxycarbonyl amino-3,3-dimethylbutanoic acid, boc-l-alpha-t-butylglycine, boc-alpha-t-butylglycine, boc-tbu-gly-oh, boc-l-tle-oh, n-tert-butoxycarbonyl-l-tert-leucine | |
(2S)-2-{[(tert-butoxy)carbonyl]amino}-3,3-dimethylbutanoic acid |
Safety and Handling
TSCA : No