missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-tert-Butyl-2-benzothiazolesulfenamide, 97%, Thermo Scientific Chemicals
$53.49 - $153.96
Chemical Identifiers
CAS | 95-31-8 |
---|---|
Molecular Formula | C11H14N2S2 |
Molecular Weight (g/mol) | 238.37 |
MDL Number | MFCD00022873 |
InChI Key | IUJLOAKJZQBENM-UHFFFAOYSA-N |
Synonym | n-tert-butyl-2-benzothiazolesulfenamide, santocure ns, nocceler ns, vulkacit nz, accel bns, pennac tbbs, vanax ns, s-benzo d thiazol-2-yl-n-tert-butyl thiohydroxylamine, 2-benzothiazolesulfenamide, n-1,1-dimethylethyl, 2-benzothiazolesulfenamide, n-tert-butyl |
PubChem CID | 7230 |
IUPAC Name | N-(1,3-benzothiazol-2-ylsulfanyl)-2-methylpropan-2-amine |
SMILES | CC(C)(C)NSC1=NC2=CC=CC=C2S1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1335618
|
Thermo Scientific Chemicals
L1335618 |
50 g |
Each for $53.49
|
|
|||||
AAL1335630
|
Thermo Scientific Chemicals
L1335630 |
250 g |
Each for $153.96
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
95-31-8 | |
238.37 | |
IUJLOAKJZQBENM-UHFFFAOYSA-N | |
7230 | |
CC(C)(C)NSC1=NC2=CC=CC=C2S1 |
C11H14N2S2 | |
MFCD00022873 | |
n-tert-butyl-2-benzothiazolesulfenamide, santocure ns, nocceler ns, vulkacit nz, accel bns, pennac tbbs, vanax ns, s-benzo d thiazol-2-yl-n-tert-butyl thiohydroxylamine, 2-benzothiazolesulfenamide, n-1,1-dimethylethyl, 2-benzothiazolesulfenamide, n-tert-butyl | |
N-(1,3-benzothiazol-2-ylsulfanyl)-2-methylpropan-2-amine |
Specifications
95-31-8 | |
1.29 | |
C11H14N2S2 | |
50 g | |
n-tert-butyl-2-benzothiazolesulfenamide, santocure ns, nocceler ns, vulkacit nz, accel bns, pennac tbbs, vanax ns, s-benzo d thiazol-2-yl-n-tert-butyl thiohydroxylamine, 2-benzothiazolesulfenamide, n-1,1-dimethylethyl, 2-benzothiazolesulfenamide, n-tert-butyl | |
CC(C)(C)NSC1=NC2=CC=CC=C2S1 | |
238.37 | |
238.38 | |
N-tert-Butyl-2-benzothiazolesulfenamide |
105°C to 110°C | |
165°C (329°F) | |
MFCD00022873 | |
158370 | |
IUJLOAKJZQBENM-UHFFFAOYSA-N | |
N-(1,3-benzothiazol-2-ylsulfanyl)-2-methylpropan-2-amine | |
7230 | |
97% |
Safety and Handling
GHS H Statement
H317
May cause an allergic skin reaction.
P261-P272-P280g-P302+P352-P333+P313-P363-P501c
H317
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 202-409-1
RTECSNumber : DL6200000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only