Learn More
N-tert-Butylisopropylamine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1722114
Description
N-tert-Butylisopropylamine, is used as a participates in the synthesis of hindered enamines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-tert-Butylisopropylamine, is used as a participates in the synthesis of hindered enamines.
Solubility
It has optimal solubility in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from oxidizing agents.
Specifications
N-tert-Butylisopropylamine | |
0.727 | |
−3°C (26°F) | |
1.398 | |
25 g | |
1846752 | |
It has optimal solubility in water. | |
CC(C)[NH2+]C(C)(C)C | |
116.23 | |
115.22 |
7515-80-2 | |
98°C | |
C7H18N | |
MFCD00037059 | |
UN2733 | |
n-tert-butylisopropylamine, 2-propanamine, 2-methyl-n-1-methylethyl, tert-butylisopropylamine, n-isopropyl-2-methylpropan-2-amine, ethanamine, 1,1-dimethyl, n-1-methylethyl, n-isopropyl-tert-butylamine, tert-butyl methylethyl amine, tert-butyl isopropyl amine | |
ZWXQPERWRDHCMZ-UHFFFAOYSA-O | |
2-methyl-N-propan-2-ylpropan-2-amine | |
82023 | |
98% |
Chemical Identifiers
7515-80-2 | |
116.23 | |
ZWXQPERWRDHCMZ-UHFFFAOYSA-O | |
82023 | |
CC(C)[NH2+]C(C)(C)C |
C7H18N | |
MFCD00037059 | |
n-tert-butylisopropylamine, 2-propanamine, 2-methyl-n-1-methylethyl, tert-butylisopropylamine, n-isopropyl-2-methylpropan-2-amine, ethanamine, 1,1-dimethyl, n-1-methylethyl, n-isopropyl-tert-butylamine, tert-butyl methylethyl amine, tert-butyl isopropyl amine | |
2-methyl-N-propan-2-ylpropan-2-amine |
Safety and Handling
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H225-H314
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber : 231-370-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only