missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
n-Triacontane, 98+%, Thermo Scientific Chemicals
$48.67 - $367.14
Chemical Identifiers
CAS | 638-68-6 |
---|---|
Molecular Formula | C30H62 |
Molecular Weight (g/mol) | 422.826 |
MDL Number | MFCD00009410 |
InChI Key | JXTPJDDICSTXJX-UHFFFAOYSA-N |
Synonym | n-triacontane, triacontane, analytical standard, triacontane, acmc-1b4uf, n-triacontane 250mg, ch3-ch2 28-ch3 |
PubChem CID | 12535 |
ChEBI | CHEBI:31006 |
IUPAC Name | triacontane |
SMILES | CCCCCCCCCCCCCCCCCCCCCCCCCCCCCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL05386MD
|
Thermo Scientific Chemicals
L05386MD |
250 mg |
Each for $48.67
|
|
|||||
AAL0538603
|
Thermo Scientific Chemicals
L0538603 |
1 g |
Each for $108.55
|
|
|||||
AAL0538606
|
Thermo Scientific Chemicals
L0538606 |
5 g |
Each for $367.14
|
|
|||||
Description
n-Triacontane is used to improve the threshold voltage and mobility for electrons of ambipolar organic light emitting field effect transistors with modified asymmetric electrodes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsn-Triacontane is used to improve the threshold voltage and mobility for electrons of ambipolar organic light emitting field effect transistors with modified asymmetric electrodes.
Solubility
Soluble in benzene, chloroform and toluene. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
638-68-6 | |
422.826 | |
JXTPJDDICSTXJX-UHFFFAOYSA-N | |
12535 | |
triacontane |
C30H62 | |
MFCD00009410 | |
n-triacontane, triacontane, analytical standard, triacontane, acmc-1b4uf, n-triacontane 250mg, ch3-ch2 28-ch3 | |
CHEBI:31006 | |
CCCCCCCCCCCCCCCCCCCCCCCCCCCCCC |
Specifications
638-68-6 | |
455°C | |
Odorless | |
MFCD00009410 | |
1777281 | |
Soluble in benzene,chloroform and toluene. Insoluble in water. | |
CCCCCCCCCCCCCCCCCCCCCCCCCCCCCC | |
422.826 | |
CHEBI:31006 | |
99% |
65°C to 68°C | |
237°C (458°F) | |
C30H62 | |
250 mg | |
n-triacontane, triacontane, analytical standard, triacontane, acmc-1b4uf, n-triacontane 250mg, ch3-ch2 28-ch3 | |
JXTPJDDICSTXJX-UHFFFAOYSA-N | |
triacontane | |
12535 | |
422.83 | |
n-Triacontane |
Safety and Handling
EINECSNumber : 211-349-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only