Learn More
n-Tridecane, 98+%, Thermo Scientific Chemicals
$74.90 - $568.48
Chemical Identifiers
CAS | 629-50-5 |
---|---|
Molecular Formula | C13H28 |
Molecular Weight (g/mol) | 184.37 |
MDL Number | MFCD00008979 |
InChI Key | IIYFAKIEWZDVMP-UHFFFAOYSA-N |
Synonym | n-tridecane, tridekan, alkanes, c12-14, dodecane, methyl, unii-a3lzf0l939, tridecane, n, trd, tridecane, analytical standard, tridecan, tridecyl group |
PubChem CID | 12388 |
ChEBI | CHEBI:35998 |
IUPAC Name | tridecane |
SMILES | CCCCCCCCCCCCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1822014
|
Thermo Scientific Chemicals
A1822014 |
25 g |
Each for $74.90
|
|
|||||
AAA1822022
|
Thermo Scientific Chemicals
A1822022 |
100 g |
Each for $153.86
|
|
|||||
AAA1822036
|
Thermo Scientific Chemicals
A1822036 |
500 g |
Each for $568.48
|
|
|||||
Description
n-Tridecane is used as a distillation chaser. It is a component of various fuels and solvents used in industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsn-Tridecane is used as a distillation chaser. It is a component of various fuels and solvents used in industry.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
629-50-5 | |
184.37 | |
IIYFAKIEWZDVMP-UHFFFAOYSA-N | |
12388 | |
tridecane |
C13H28 | |
MFCD00008979 | |
n-tridecane, tridekan, alkanes, c12-14, dodecane, methyl, unii-a3lzf0l939, tridecane, n, trd, tridecane, analytical standard, tridecan, tridecyl group | |
CHEBI:35998 | |
CCCCCCCCCCCCC |
Specifications
629-50-5 | |
0.755 | |
79°C (174°F) | |
1.425 | |
MFCD00008979 | |
1733089 | |
Immiscible with water. | |
CCCCCCCCCCCCC | |
184.37 | |
CHEBI:35998 | |
≥98% |
-5°C | |
235°C | |
C13H28 | |
CH3(CH2)11CH3 | |
25 g | |
n-tridecane, tridekan, alkanes, c12-14, dodecane, methyl, unii-a3lzf0l939, tridecane, n, trd, tridecane, analytical standard, tridecan, tridecyl group | |
IIYFAKIEWZDVMP-UHFFFAOYSA-N | |
tridecane | |
12388 | |
184.37 | |
n-Tridecane |
Safety and Handling
GHS H Statement
H304
May be fatal if swallowed and enters airways.
P210-P280-P301+P310-P331-P370+P378q-P501c
H227-H304
EINECSNumber : 211-093-4
RTECSNumber : YD3025000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only