missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-(α,α,α-Trifluoro-p-tolyl)piperazine, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 367120050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
N-(α, α, α-Trifluoro-p-tolyl)piperazine | |
30459-17-7 | |
100.0 | |
97.5% min. (GC) | |
C11H13F3N3 | |
5g | |
IBQMAPSJLHRQPE-UHFFFAOYSA-N | |
1-[4-(trifluoromethyl)phenyl]piperazine | |
121718 | |
98% |
98% | |
97.5 | |
Authentic | |
Glass bottle | |
MFCD00040765 | |
1-4-trifluoromethylphenyl piperazine, 1-4-trifluoromethyl phenyl piperazine, piperazine, 1-4-trifluoromethyl phenyl, 1-4-trifluoromethylphenyl-piperazine, 1-4-trifluoromethyl-phenyl-piperazine, n-4-trifluoromethylphenyl piperazine, 4-trifluoromethyl phenyl piperazine, zlchem 639, acmc-209hfy | |
C1CN(CCN1)C2=CC=C(C=C2)C(F)(F)F | |
230.23 | |
230.23 |
Chemical Identifiers
30459-17-7 | |
230.23 | |
IBQMAPSJLHRQPE-UHFFFAOYSA-N | |
121718 | |
C1CN(CCN1)C2=CC=C(C=C2)C(F)(F)F |
C11H13F3N3 | |
MFCD00040765 | |
1-4-trifluoromethylphenyl piperazine, 1-4-trifluoromethyl phenyl piperazine, piperazine, 1-4-trifluoromethyl phenyl, 1-4-trifluoromethylphenyl-piperazine, 1-4-trifluoromethyl-phenyl-piperazine, n-4-trifluoromethylphenyl piperazine, 4-trifluoromethyl phenyl piperazine, zlchem 639, acmc-209hfy | |
1-[4-(trifluoromethyl)phenyl]piperazine |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 250-210-3
RUO â Research Use Only