missing translation for 'onlineSavingsMsg'
Learn More

N-(Triphenylmethyl)-DL-serine Methyl Ester 98.0+%, TCI America™

$52.19 - $260.97

Chemical Identifiers

CAS 13515-76-9
Molecular Formula C23H23NO3
Molecular Weight (g/mol) 361.441
InChI Key LXAWQKKSNNYYEK-UHFFFAOYSA-N
Synonym N-Trityl-DL-serine Methyl Ester, Trt-DL-Ser-OMe
PubChem CID 279018
IUPAC Name methyl 3-hydroxy-2-(tritylamino)propanoate
SMILES COC(=O)C(CO)NC(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T25005G
View Documents
TCI America
T25005G
5 g N/A
Only null left
 
T250025G
View Documents
TCI America
T250025G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

13515-76-9
361.441
N-Trityl-DL-serine Methyl Ester, Trt-DL-Ser-OMe
methyl 3-hydroxy-2-(tritylamino)propanoate
C23H23NO3
LXAWQKKSNNYYEK-UHFFFAOYSA-N
279018
COC(=O)C(CO)NC(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

13515-76-9
White
5 g
LXAWQKKSNNYYEK-UHFFFAOYSA-N
methyl 3-hydroxy-2-(tritylamino)propanoate
279018
≥98.0% (GC,T)
N-(Triphenylmethyl)-DL-serine Methyl Ester
146°C
C23H23NO3
N-Trityl-DL-serine Methyl Ester, Trt-DL-Ser-OMe
COC(=O)C(CO)NC(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3
361.441
361.44
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'