Learn More
n-Undecane, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 140660025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
n-Undecane | |
1120-21-4 | |
100.0 | |
Colorless | |
196.0°C | |
Authentic | |
Glass bottle | |
1.4160 to 1.4180 | |
MFCD00008959 | |
01, IV, 487 | |
n-undecane, hendecane, n-hendecane, decane, methyl, unii-jv0qt00nue, ccris 3796, jv0qt00nue, undecane, undecane, analytical standard, und | |
CCCCCCCCCCC | |
156.31 | |
CHEBI:46342 | |
0.6hPa at 20°C | |
Liquid |
99% | |
98.5 | |
-26.0°C | |
0.7400g/mL | |
65°C | |
99% | |
C11H24 | |
CH3(CH2)9CH3 | |
2.5 L | |
0.74 | |
RSJKGSCJYJTIGS-UHFFFAOYSA-N | |
undecane | |
14257 | |
156.31 | |
99% |
Chemical Identifiers
1120-21-4 | |
156.31 | |
RSJKGSCJYJTIGS-UHFFFAOYSA-N | |
14257 | |
undecane |
C11H24 | |
MFCD00008959 | |
n-undecane, hendecane, n-hendecane, decane, methyl, unii-jv0qt00nue, ccris 3796, jv0qt00nue, undecane, undecane, analytical standard, und | |
CHEBI:46342 | |
CCCCCCCCCCC |
Safety and Handling
GHS H Statement:
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
EINECSNumber : 214-300-6
RTECSNumber : YQ1525000
TSCA : TSCA
RUO – Research Use Only