missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Undecane, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 140661000

 View more versions of this product

Catalog No. AC140661000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

n-Undecane
1120-21-4
100.0
0.7400g/mL
99%
1.4160 to 1.4180 (20°C, 589nm)
MFCD00008959
01, IV, 487
RSJKGSCJYJTIGS-UHFFFAOYSA-N
undecane
14257
156.31
≥98.5%
99%
98.5
Colorless
Authentic
C11H24
CH3(CH2)9CH3
100 mL
n-undecane, hendecane, n-hendecane, decane, methyl, unii-jv0qt00nue, ccris 3796, jv0qt00nue, undecane, undecane, analytical standard, und
CCCCCCCCCCC
156.31
CHEBI:46342
0.6hPa at 20°C
Liquid

Chemical Identifiers

1120-21-4
156.31
RSJKGSCJYJTIGS-UHFFFAOYSA-N
14257
undecane
C11H24
MFCD00008959
n-undecane, hendecane, n-hendecane, decane, methyl, unii-jv0qt00nue, ccris 3796, jv0qt00nue, undecane, undecane, analytical standard, und
CHEBI:46342
CCCCCCCCCCC
Safety and Handling

Safety and Handling

GHS H Statement:
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 214-300-6

RTECSNumber : YQ1525000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only