missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Vinyl-2-pyrrolidone, 99%, stabilized with NaOH, Thermo Scientific Chemicals

$98.18 - $480.06

Chemical Identifiers

CAS 88-12-0
Molecular Formula C6H9NO
Molecular Weight (g/mol) 111.14
MDL Number MFCD00003197
InChI Key WHNWPMSKXPGLAX-UHFFFAOYSA-N
Synonym n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone
PubChem CID 6917
ChEBI CHEBI:82551
IUPAC Name 1-ethenylpyrrolidin-2-one
SMILES C=CN1CCCC1=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC140920050
SDS View Documents
Thermo Scientific Chemicals
140920050
5 mL
Each for $98.18
 
AC140925000
SDS View Documents
Thermo Scientific Chemicals
140925000
500 mL
Each for $144.15
 
AC140920025
SDS View Documents
Thermo Scientific Chemicals
140920025
2.5 L
Each for $480.06
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Chemical Identifiers

88-12-0
111.14
WHNWPMSKXPGLAX-UHFFFAOYSA-N
6917
1-ethenylpyrrolidin-2-one
C6H9NO
MFCD00003197
n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone
CHEBI:82551
C=CN1CCCC1=O

Specifications

88-12-0
100.0
Colorless to Yellow
218°C
Authentic
Glass bottle
1.5119 to 1.5139
5 mL
15,7814
Solubility in water: 52.1g/L (25°C). Other solubilities: miscible with acetone,diethyl ether,ethanol,,toluene,benzene,very good soluble in organic solvents
C=CN1CCCC1=O
111.14
6917
CHEBI:82551
99%
N-Vinyl-2-pyrrolidone, 98%
98.5
13°C to 14°C
1.0430g/mL
95°C
0.2% max. (Pyrrolidone) (GC)
C6H9NO
MFCD00003197
1.043
n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone
WHNWPMSKXPGLAX-UHFFFAOYSA-N
1-ethenylpyrrolidin-2-one
0.1% flaked caustic soda
2.4 mPa.s (20°C)
111.14
Liquid
Safety and Handling

Safety and Handling

GHS H Statement
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Danger

EINECSNumber : 201-800-4

RTECSNumber : UY6107000

TSCA : TSCA

Recommended Storage : Product may darken on storage

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only