missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Vinylphthalimide 98.0+%, TCI America™
$226.65 - $762.97
Chemical Identifiers
CAS | 3485-84-5 |
---|---|
Molecular Formula | C10H7NO2 |
Molecular Weight (g/mol) | 173.17 |
MDL Number | MFCD00078446 |
InChI Key | IGDLZDCWMRPMGL-UHFFFAOYSA-N |
Synonym | n-vinylphthalimide, 2-vinylisoindoline-1,3-dione, phthalimide, n-vinyl, 1h-isoindole-1,3 2h-dione, 2-ethenyl, 2-vinyl-1h-isoindole-1,3 2h-dione, 2-ethenyl-1h-isoindole-1,3 2h-dione, 2-ethenyl-1,3-dihydro-1,3-dioxoisoindole, 2-ethenyl-2,3-dihydro-1h-isoindole-1,3-dione, 2-vinylbenzo c azolidine-1,3-dione, n-vinylphtalimide |
PubChem CID | 77035 |
IUPAC Name | 2-ethenyl-2,3-dihydro-1H-isoindole-1,3-dione |
SMILES | C=CN1C(=O)C2=CC=CC=C2C1=O |
Chemical Identifiers
3485-84-5 | |
173.17 | |
IGDLZDCWMRPMGL-UHFFFAOYSA-N | |
77035 | |
C=CN1C(=O)C2=CC=CC=C2C1=O |
C10H7NO2 | |
MFCD00078446 | |
n-vinylphthalimide, 2-vinylisoindoline-1,3-dione, phthalimide, n-vinyl, 1h-isoindole-1,3 2h-dione, 2-ethenyl, 2-vinyl-1h-isoindole-1,3 2h-dione, 2-ethenyl-1h-isoindole-1,3 2h-dione, 2-ethenyl-1,3-dihydro-1,3-dioxoisoindole, 2-ethenyl-2,3-dihydro-1h-isoindole-1,3-dione, 2-vinylbenzo c azolidine-1,3-dione, n-vinylphtalimide | |
2-ethenyl-2,3-dihydro-1H-isoindole-1,3-dione |
Specifications
3485-84-5 | |
151°C | |
MFCD00078446 | |
n-vinylphthalimide, 2-vinylisoindoline-1,3-dione, phthalimide, n-vinyl, 1h-isoindole-1,3 2h-dione, 2-ethenyl, 2-vinyl-1h-isoindole-1,3 2h-dione, 2-ethenyl-1h-isoindole-1,3 2h-dione, 2-ethenyl-1,3-dihydro-1,3-dioxoisoindole, 2-ethenyl-2,3-dihydro-1h-isoindole-1,3-dione, 2-vinylbenzo c azolidine-1,3-dione, n-vinylphtalimide | |
C=CN1C(=O)C2=CC=CC=C2C1=O | |
173.17 | |
173.17 | |
Crystalline Powder |
83°C | |
C10H7NO2 | |
5 g | |
IGDLZDCWMRPMGL-UHFFFAOYSA-N | |
2-ethenyl-2,3-dihydro-1H-isoindole-1,3-dione | |
77035 | |
≥98.0% (GC) | |
N-Vinylphthalimide |
Safety and Handling
TSCA : Yes