missing translation for 'onlineSavingsMsg'
Learn More

N1-(3-acetyl-4-hydroxyphenyl)acetamide, Thermo Scientific™

Chemical Identifiers

CAS 7298-67-1
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.202
InChI Key DIQSYMRVTOVKQT-UHFFFAOYSA-N
Synonym n-3-acetyl-4-hydroxyphenyl acetamide, n1-3-acetyl-4-hydroxyphenyl acetamide, n-3-acetyl-4-hydroxy-phenyl acetamide, 5'-acetamido-2'-hydroxyacetophenone, acetamide, n-3-acetyl-4-hydroxyphenyl, ksc380g7h, 3'-acetyl-4'-hydroxyacetanilide, 5-acetamido-2-hydroxyacetophenone, 5-acetamido-2-hydroxy-acetophenone, n-3-acetyl-4-hydroxyphenyl-acetamide
PubChem CID 81720
IUPAC Name N-(3-acetyl-4-hydroxyphenyl)acetamide
SMILES CC(=O)C1=C(C=CC(=C1)NC(=O)C)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
BTB10078DA
SDS View Documents
Thermo Scientific Chemicals
BTB10078DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTB10078DE
SDS View Documents
Thermo Scientific Chemicals
BTB10078DE
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

7298-67-1
193.202
n-3-acetyl-4-hydroxyphenyl acetamide, n1-3-acetyl-4-hydroxyphenyl acetamide, n-3-acetyl-4-hydroxy-phenyl acetamide, 5'-acetamido-2'-hydroxyacetophenone, acetamide, n-3-acetyl-4-hydroxyphenyl, ksc380g7h, 3'-acetyl-4'-hydroxyacetanilide, 5-acetamido-2-hydroxyacetophenone, 5-acetamido-2-hydroxy-acetophenone, n-3-acetyl-4-hydroxyphenyl-acetamide
N-(3-acetyl-4-hydroxyphenyl)acetamide
C10H11NO3
DIQSYMRVTOVKQT-UHFFFAOYSA-N
81720
CC(=O)C1=C(C=CC(=C1)NC(=O)C)O

Specifications

7298-67-1
C10H11NO3
n-3-acetyl-4-hydroxyphenyl acetamide, n1-3-acetyl-4-hydroxyphenyl acetamide, n-3-acetyl-4-hydroxy-phenyl acetamide, 5'-acetamido-2'-hydroxyacetophenone, acetamide, n-3-acetyl-4-hydroxyphenyl, ksc380g7h, 3'-acetyl-4'-hydroxyacetanilide, 5-acetamido-2-hydroxyacetophenone, 5-acetamido-2-hydroxy-acetophenone, n-3-acetyl-4-hydroxyphenyl-acetamide
CC(=O)C1=C(C=CC(=C1)NC(=O)C)O
193.202
193.2
N1-(3-Acetyl-4-hydroxyphenyl)acetamide
97%
1g
DIQSYMRVTOVKQT-UHFFFAOYSA-N
N-(3-acetyl-4-hydroxyphenyl)acetamide
81720
97%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'