missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N2-Isobutyrylguanosine Monohydrate, TCI America™

$135.75 - $867.71

Chemical Identifiers

CAS 64350-24-9
Molecular Formula C14H21N5O7
Molecular Weight (g/mol) 371.35
MDL Number MFCD00274102,MFCD00274102
InChI Key JXIMAIWXKDBJKP-UHFFFAOYNA-N
PubChem CID 56924510
IUPAC Name N-{9-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-6-oxo-6,9-dihydro-3H-purin-2-yl}-2-methylpropanamide hydrate
SMILES O.CC(C)C(=O)NC1=NC(=O)C2=C(N1)N(C=N2)C1OC(CO)C(O)C1O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I0699100MG
View Documents
TCI America
I0699100MG
100 mg
Each for $135.75
 
I06991G
View Documents
TCI America
I06991G
1 g
Each for $867.71
 
Description

Description

Specifications

Chemical Identifiers

64350-24-9
371.35
JXIMAIWXKDBJKP-UHFFFAOYNA-N
N-{9-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-6-oxo-6,9-dihydro-3H-purin-2-yl}-2-methylpropanamide hydrate
C14H21N5O7
MFCD00274102,MFCD00274102
56924510
O.CC(C)C(=O)NC1=NC(=O)C2=C(N1)N(C=N2)C1OC(CO)C(O)C1O

Specifications

64350-24-9
White
MFCD00274102,MFCD00274102
JXIMAIWXKDBJKP-UHFFFAOYNA-N
N-{9-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-6-oxo-6,9-dihydro-3H-purin-2-yl}-2-methylpropanamide hydrate
56924510
Crystalline Powder
148°C
C14H21N5O7
100 mg
O.CC(C)C(=O)NC1=NC(=O)C2=C(N1)N(C=N2)C1OC(CO)C(O)C1O
371.35
353.34
N2-Isobutyrylguanosine Monohydrate
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'