Thermo Scientific™ Nalgene™ 870 PFA Tubing
Utilize translucent 870 tubing for transferring aggressive and corrosive chemical fluids. Useful over a very wide temperature range (-270° to +260°C) with low water absorption.
$617.44 - $4188.47
Specifications
Certifications/Compliance Food-Grade materials comply with 21 CFR 177.1520 for food packaging
Autoclavable Autoclavable
Material PFA
Packaging Quantity 25
Color Transparent
View More Specs
Products 5
Catalog Number Mfr. No. Diameter (English) Inner Diameter (English) Outer Maximum Pressure Price Quantity  
Catalog Number Mfr. No. Diameter (English) Inner Diameter (English) Outer Maximum Pressure Price Quantity  
14-176-260
View Documents
Thermo Scientific™
80510125
1/16 in. 1/8 in. 478 psig at 73°F (23°C)
Pack of 25 Each for $617.44
Case of 2 Pack for $1,234.88
14-176-259
View Documents
Thermo Scientific™
80510187
1/8 in. 3/16 in. 274 psig at 73°F (23°C), 137psig at 185°F (85°C)
Pack of 25 Each for $939.56
Case of 2 Pack for $1,879.12
14-176-258
View Documents
Thermo Scientific™
80510250
3/64 in. 1/4 in. 290 psig at 73°F (23°C), 145psig at 185°F (85°C)
Pack of 25 Each for $1,828.63
Case of 2 Pack for $3,657.26
14-176-256
View Documents
Thermo Scientific™
80510375
1/4 in. 3/8 in. 248 psig at 73°F (23°C), 124psig at 185°F (85°C)
Case of 25 Each for $3,181.72
14-176-254
View Documents
Thermo Scientific™
80510500
3/8 in. 1/2 in. 85 psig at 185°F (85°C)
Case of 25 Each for $4,188.47
Description
Description
Utilize translucent Thermo Scientific™ Nalgene™ 870 Tubing for transferring aggressive and corrosive chemical fluids. Useful over a very wide temperature range (-270° to +260°C) with low water absorption. Excellent for low-pressure use with mechanical, compression fittings at room tempearature. Food-grade materials comply with 21 CFR 177.1520 for food packaging.
Features:
  • Wider temperature range -270° to +260°C (-518° to +500°F)
  • High level of inertness for high purity applications
  • Sterilization by autoclaving, gas, dry heat and disinfectants
  • Two 25′ coils per case except larger sizes
Note:
Do not sterilize by radiation. Should not be used with molten alkali metals, elemental fluorine and fluorine precursors at elevated temperatures or concentrated perchloric acid.
Please recycle the corrugate cartons in which your Nalgene  products are packaged.
.
Specifications
Specifications
Food-Grade materials comply with 21 CFR 177.1520 for food packaging
PFA
Transparent
10 x OD bend radius
260°C
Autoclavable
25
60(Shore D)
500°F
Semi-rigid Tubing
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'