missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Nalgene™ Büchner Two-Piece Polypropylene Funnels

Separate the lightweight and durable two-piece polypropylene funnels for easy cleaning. These funnels won't chip, crack or break.

Supplier:  Thermo Scientific™ 42801100

 View more versions of this product

Catalog No. 10-362E



Explore available promotions

Description

Description

Separate the lightweight and durable Thermo Scientific™ Nalgene™ Büchner Two-Piece Polypropylene Funnels for easy cleaning. These autoclavable funnels won't chip, crack or break making them a valuble asset to lab safety programs.

  • Sturdy funnels consist of cup and funnel with perforated plate for easy cleaning and good chemical resistance
  • All sizes are made entirely of polypropylene and are autoclavable to 135°C (275°F)
Specifications

Specifications

132 mm
Polypropylene
Autoclavable
92 mm
919 mL
214 mm
Funnel Buchner PP 110mm
1/Pk.; 4/Cs.
Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions