missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Biohazardous Waste Containers
Conduct secondary containment of biohazardous waste with these biohazardous waste containers, which feature convenient one-step sterilization and enable one-handed disposal of materials.
$734.83 - $1,047.79
Specifications
Certifications/Compliance | U.S. OSHA Standard 29 CFR Part 1910.1030 |
---|---|
Material | Polypropylene |
Sterility | Non-sterile |
Type | Waste Container Biohazardous |
No. per Case | 1 |
Catalog Number | Mfr. No. | Capacity (Metric) | Diameter (Metric) Outer | Height (Metric) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Diameter (Metric) Outer | Height (Metric) | Price | Quantity | |||
11-815-20
|
Thermo Scientific™
63700004 |
5.5 L | 22 cm | 28 cm |
N/A
|
|
|||
14-293-63A
|
Thermo Scientific™
63700005 |
19 L | 28 cm | 43 cm |
N/A
|
|
|||
14-293-63B
|
Thermo Scientific™
63700015 |
57 L | 34 cm | 74 cm |
N/A
|
|
|||
Description
Conduct secondary containment of biohazardous waste with Thermo Scientific™ Nalgene™ Biohazardous Waste Containers, which feature convenient one-step sterilization—no need to remove the bag. A port opening in the cover allows for one-handed disposal of materials, limiting exposure to infectious waste. The containers hold standard sized autoclave bags (not included).
- Polypropylene copolymer (PPCO) components will not rust, dent or puncture like other materials
- Complies with U.S. OSHA Standard 29 CFR Part 1910.1030 for use as protection against bloodborne pathogens
- Autoclavable/Biohazard
- Symbol printed in English and Spanish
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in.
.
Specifications
U.S. OSHA Standard 29 CFR Part 1910.1030 | |
Non-sterile | |
1 |
Polypropylene | |
Waste Container Biohazardous | |
1 |