missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Nalgene™ Double-Scale PMP Pharmaceutical Graduates

Avoid breakage with transparent and shatterproof double-scale PMP pharmaceutical graduates. Lighter in weight than glass, they provide excellent chemical resistance.

Supplier:  Thermo Scientific™ 36730016

 View more versions of this product

Catalog No. 11-423C



Explore available promotions

Description

Description

Avoid breakage with transparent and shatterproof Thermo Scientific™ Nalgene™ Double-Scale PMP Pharmaceutical Graduates. Lighter in weight than glass, they provide excellent chemical resistance. They are graduated in ounces and milliliters with no meniscus to confuse readings.

  • Meet the requirement of ASTM E1094
  • Autoclavable (will affect accuracy)
  • Graduated
Specifications

Specifications

ASTM E1094
Dual Scale mL and ounces
±4.8 mL
1
16 oz., 500 mL
Graduated Cylinder
PMP
500 mL
16 oz.
8
Graduated Cylinder
Graduated Cylinder Pharmaceutical PMP 500mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions