Learn More
Thermo Scientific™ CryoTube™ Holders
Increase the functionality of your cryogenic tubes with these benchtop tube and vial holders in a variety of compartment configurations with single-handed capping and decapping.
$161.70 - $235.30
| Catalog Number | Mfr. No. | Holds | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Holds | Price | Quantity | |||||
|
03-337-7E
|
Thermo Scientific™
50300510 |
50 Cryogenic Tubes |
Each for $161.70
Case of 4 Each for $646.80
|
|
|||||
|
03-337-7Z
|
Thermo Scientific™
50300505 |
25 Cryogenic Tubes |
Each for $219.57
Case of 4 Each for $878.28
|
|
|||||
|
12-565-178
|
Thermo Scientific™
376589 |
40 Cryogenic Tubes |
Each for $235.30
Case of 15 Each for $3,529.50
|
|
|||||
Description
Expand the functionality of cryogenic tube products with Thermo Scientific™ CryoTube™ Holders — benchtop tube and vial containers for single-handed capping and decapping. The product range includes Nalgene™ and Nunc™ containers in various sizes and compartment configurations.
- Polystyrene coders snap into the caps of Nalgene Cryogenic Tubes for easy sample identification
- Polycarbonate Benchtop Cryogenic Tube Holder has anti-rotation feature for one handed capping and decapping
Specifications
| Vial Holder |