missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Nalgene™ Large Narrow-Mouth LDPE Bottles

Collect and store distilled water in bottles that offer flexibility, impact resistance and visibility of contents.

Supplier:  Thermo Scientific™ 22020010

 View more versions of this product

Catalog No. 02-923J

Explore available promotions

Description

Description

Store chemicals and standards and collect and store distilled water. Thermo Scientific™ Nalgene™ Large Narrow-Mouth LDPE Bottles are translucent and offer flexibility, impact resistance and visibility of contents. The extremely low trace metal content in low-density polyethylene makes them ideal for trace metal analysis. These bottles provide excellent chemical resistance to most acids, bases and alcohols. Made only from high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables.

Features:
  • Excellent chemical resistance to most acids, bases and alcohols make these versatile bottles
  • Can be used in freezer to -100°C (-148°F) for long-term storage
  • 4 and 8L sizes feature a built-in shoulder loop on larger sizes for attaching identification tag
  • Translucence allows for easy viewing of liquid levels
  • Linerless caps provide the ultimate in leakproof† protection without the use of liner that can wrinkle, cause leaks or contiminate your reagents

Includes:

Leakproof† polypropylene screw caps

Note:

Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided.

Specifications

Specifications

1 gal., 4 L
1 gal.
Large Narrow-Mouth Bottle
LDPE
1
White
Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
No
Round
Narrow Mouth Bottle
13.1 in.
Polypropylene
152 mm
No
4 L
Translucent
25 mm
6
Non-sterile
38 mm
Yes
Narrow
333 mm
6 in.
Unlined
38-430 mm
Screw Cap
Videos
SDS
Documents

Documents

Promotions

Promotions

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.